missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Amino-5-methylpyrazine, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals H61286MD

 View more versions of this product

Catalog No. AAH61286MD


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

2-Amino-5-methylpyrazine
C5H7N3
250 mg
ZNQOALAKPLGUPH-UHFFFAOYSA-N
5-methylpyrazin-2-amine
313215
98%
5521-58-4
MFCD08437664
2-amino-5-methylpyrazine, 5-methyl-pyrazin-2-ylamine, 5-methyl-2-pyrazinamine, pyrazinamine, 5-methyl, 2-amino-5 methylpyrazine, 5-methyl-2-aminopyrazine, 2-amino-5-methyl pyrazine, 5-methylpyrazine-2-ylamine, 2-pyrazinamine, 5-methyl, pubchem15710
CC1=CN=C(C=N1)N
109.132
109.13
Solid

Chemical Identifiers

5521-58-4
109.132
ZNQOALAKPLGUPH-UHFFFAOYSA-N
313215
CC1=CN=C(C=N1)N
C5H7N3
MFCD08437664
2-amino-5-methylpyrazine, 5-methyl-pyrazin-2-ylamine, 5-methyl-2-pyrazinamine, pyrazinamine, 5-methyl, 2-amino-5 methylpyrazine, 5-methyl-2-aminopyrazine, 2-amino-5-methyl pyrazine, 5-methylpyrazine-2-ylamine, 2-pyrazinamine, 5-methyl, pubchem15710
5-methylpyrazin-2-amine
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only