missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2-Amino-5-nitrobenzonitrile 98.0+%, TCI America™
$120.48 - $1145.51
Chemical Identifiers
CAS | 17420-30-3 |
---|---|
Molecular Formula | C7H5N3O2 |
Molecular Weight (g/mol) | 163.136 |
MDL Number | MFCD00007362 |
InChI Key | MGCGMYPNXAFGFA-UHFFFAOYSA-N |
Synonym | 5-nitroanthranilonitrile, 2-cyano-4-nitroaniline, benzonitrile, 2-amino-5-nitro, 2-kyan-4-nitroanilin, unii-sez79wra1j, 2-amino-5-nitro-benzonitrile, ccris 9092, 2-kyan-4-nitroanilin czech, aniline, 2-cyano-4-nitro, sez79wra1j |
PubChem CID | 28532 |
IUPAC Name | 2-amino-5-nitrobenzonitrile |
SMILES | C1=CC(=C(C=C1[N+](=O)[O-])C#N)N |
Chemical Identifiers
17420-30-3 | |
163.136 | |
MGCGMYPNXAFGFA-UHFFFAOYSA-N | |
28532 | |
C1=CC(=C(C=C1[N+](=O)[O-])C#N)N |
C7H5N3O2 | |
MFCD00007362 | |
5-nitroanthranilonitrile, 2-cyano-4-nitroaniline, benzonitrile, 2-amino-5-nitro, 2-kyan-4-nitroanilin, unii-sez79wra1j, 2-amino-5-nitro-benzonitrile, ccris 9092, 2-kyan-4-nitroanilin czech, aniline, 2-cyano-4-nitro, sez79wra1j | |
2-amino-5-nitrobenzonitrile |
Specifications
17420-30-3 | |
C7H5N3O2 | |
25 g | |
MGCGMYPNXAFGFA-UHFFFAOYSA-N | |
2-amino-5-nitrobenzonitrile | |
28532 | |
≥98.0% (GC) | |
2-Amino-5-nitrobenzonitrile |
210°C | |
MFCD00007362 | |
5-nitroanthranilonitrile, 2-cyano-4-nitroaniline, benzonitrile, 2-amino-5-nitro, 2-kyan-4-nitroanilin, unii-sez79wra1j, 2-amino-5-nitro-benzonitrile, ccris 9092, 2-kyan-4-nitroanilin czech, aniline, 2-cyano-4-nitro, sez79wra1j | |
C1=CC(=C(C=C1[N+](=O)[O-])C#N)N | |
163.136 | |
163.14 | |
Crystalline Powder |
Safety and Handling
EINECSNumber : (3)-1806
RTECSNumber : BX2500000
TSCA : Yes