Learn More
2-Amino-5-nitrobenzophenone, 98%, Thermo Scientific Chemicals
$40.18 - $266.62
Chemical Identifiers
CAS | 1775-95-7 |
---|---|
Molecular Formula | C13H10N2O3 |
Molecular Weight (g/mol) | 242.23 |
MDL Number | MFCD00007364 |
InChI Key | PZPZDEIASIKHPY-UHFFFAOYSA-N |
Synonym | 2-amino-5-nitrobenzophenone, 2-amino-5-nitrophenyl phenyl methanone, methanone, 2-amino-5-nitrophenyl phenyl, 2-amino-5-nitrophenyl phenylmethanone, 2-amino-5-nitrophenyl-phenylmethanone, 2-benzoyl-4-nitroaniline, 5-nitro-2-aminobenzophenone, 2-amino-5-nitro-phenyl-phenyl-methanone, 2-amino-5-nitrophenyl phenyl ketone, pubchem3254 |
PubChem CID | 15681 |
IUPAC Name | 2-benzoyl-4-nitroaniline |
SMILES | NC1=CC=C(C=C1C(=O)C1=CC=CC=C1)[N+]([O-])=O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAB2377306
|
Thermo Scientific Chemicals
B2377306 |
5 g |
Each for $40.18
|
|
|||||
AAB2377314
|
Thermo Scientific Chemicals
B2377314 |
25 g |
Each for $109.58
|
|
|||||
AAB2377322
|
Thermo Scientific Chemicals
B2377322 |
100 g |
Each for $266.62
|
|
|||||
Description
It is used in the determination of nitrazepam in serum by gas-liquid chromatography; application in bioavailability studies and in second-harmonic generation in aromatic organic compounds.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is used in the determination of nitrazepam in serum by gas-liquid chromatography; application in bioavailability studies and in second-harmonic generation in aromatic organic compounds.
Solubility
Soluble in chloroform, dichloromethane, dimethyl sulfoxide, ethyl acetate and methanol. Soluble in water 3 mg/L (20°C).
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Chemical Identifiers
1775-95-7 | |
242.23 | |
PZPZDEIASIKHPY-UHFFFAOYSA-N | |
15681 | |
NC1=CC=C(C=C1C(=O)C1=CC=CC=C1)[N+]([O-])=O |
C13H10N2O3 | |
MFCD00007364 | |
2-amino-5-nitrobenzophenone, 2-amino-5-nitrophenyl phenyl methanone, methanone, 2-amino-5-nitrophenyl phenyl, 2-amino-5-nitrophenyl phenylmethanone, 2-amino-5-nitrophenyl-phenylmethanone, 2-benzoyl-4-nitroaniline, 5-nitro-2-aminobenzophenone, 2-amino-5-nitro-phenyl-phenyl-methanone, 2-amino-5-nitrophenyl phenyl ketone, pubchem3254 | |
2-benzoyl-4-nitroaniline |
Specifications
1775-95-7 | |
C13H10N2O3 | |
5 g | |
Soluble in chloroform,dichloromethane,dimethyl sulfoxide,ethyl acetate and methanol. Soluble in water 3mg/L (20°C). | |
NC1=CC=C(C=C1C(=O)C1=CC=CC=C1)[N+]([O-])=O | |
242.23 | |
242.23 | |
2-Amino-5-nitrobenzophenone |
166°C to 168°C | |
MFCD00007364 | |
2-amino-5-nitrobenzophenone, 2-amino-5-nitrophenyl phenyl methanone, methanone, 2-amino-5-nitrophenyl phenyl, 2-amino-5-nitrophenyl phenylmethanone, 2-amino-5-nitrophenyl-phenylmethanone, 2-benzoyl-4-nitroaniline, 5-nitro-2-aminobenzophenone, 2-amino-5-nitro-phenyl-phenyl-methanone, 2-amino-5-nitrophenyl phenyl ketone, pubchem3254 | |
PZPZDEIASIKHPY-UHFFFAOYSA-N | |
2-benzoyl-4-nitroaniline | |
15681 | |
98% |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P264b-P270-P301+P312-P330-P501c
H302
EINECSNumber : 217-207-9
RTECSNumber : PC4935000
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only