Learn More
2-Amino-5-nitrophenol, 95%, Thermo Scientific Chemicals
$60.81 - $186.27
Chemical Identifiers
CAS | 121-88-0 |
---|---|
Molecular Formula | C6H6N2O3 |
Molecular Weight (g/mol) | 154.125 |
MDL Number | MFCD00007692 |
InChI Key | DOPJTDJKZNWLRB-UHFFFAOYSA-N |
Synonym | 2-hydroxy-4-nitroaniline, 5-nitro-2-aminophenol, phenol, 2-amino-5-nitro, ursol yellow brown a, rodol yba, 3-nitro-6-aminophenol, 3-hydroxy-4-aminonitrobenzene, 2-amino-5-nitro-phenol, ccris 754, 4-amino-3-hydroxynitrobenzene |
PubChem CID | 4984721 |
ChEBI | CHEBI:82384 |
IUPAC Name | 2-amino-5-nitrophenol |
SMILES | C1=CC(=C(C=C1[N+](=O)[O-])O)N |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
AAB2087014
|
Thermo Scientific Chemicals
B2087014 |
25 g |
Each for $60.81
|
|
||||||
AAB2087022
|
Thermo Scientific Chemicals
B2087022 |
100 g |
Each for $186.27
|
|
||||||
Description
2-Amino-5-nitrophenol has been used as starting material in the synthesis of series of (Z)-2-(substituted aryl)-N-(3-oxo-4-(substituted carbamothioyl)-3,4-dihydro-2H-benzo[b][1,4]oxazin-7-yl) hydrazine carboxamides, as semi permanent (nonoxidative) hair colorant and as toner in permanent (oxidative) hair dye products, in synthesis of series of 7-benzylamino-2H-1,4-benzoxazin-3(4H)-ones, anticonvulsant agents. Also used as a reactant for, diazotation and coupling reactions, preparation of biologically and pharmacologically active molecules. It is also applied in hair dyes and to make azo dyes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2-Amino-5-nitrophenol has been used as starting material in the synthesis of series of (Z)-2-(substituted aryl)-N-(3-oxo-4-(substituted carbamothioyl)-3,4-dihydro-2H-benzo[b][1,4]oxazin-7-yl) hydrazine carboxamides, as semi permanent (nonoxidative) hair colorant and as toner in permanent (oxidative) hair dye products, in synthesis of series of 7-benzylamino-2H-1,4-benzoxazin-3(4H)-ones, anticonvulsant agents. Also used as a reactant for, diazotation and coupling reactions, preparation of biologically and pharmacologically active molecules. It is also applied in hair dyes and to make azo dyes.
Solubility
Insoluble in water. Solubility in methanol gives very faint turbidity. Soluble in alcohol, benzene, acetone, and diethyl ether.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Chemical Identifiers
121-88-0 | |
154.125 | |
DOPJTDJKZNWLRB-UHFFFAOYSA-N | |
4984721 | |
2-amino-5-nitrophenol |
C6H6N2O3 | |
MFCD00007692 | |
2-hydroxy-4-nitroaniline, 5-nitro-2-aminophenol, phenol, 2-amino-5-nitro, ursol yellow brown a, rodol yba, 3-nitro-6-aminophenol, 3-hydroxy-4-aminonitrobenzene, 2-amino-5-nitro-phenol, ccris 754, 4-amino-3-hydroxynitrobenzene | |
CHEBI:82384 | |
C1=CC(=C(C=C1[N+](=O)[O-])O)N |
Specifications
121-88-0 | |
C6H6N2O3 | |
25 g | |
2-hydroxy-4-nitroaniline, 5-nitro-2-aminophenol, phenol, 2-amino-5-nitro, ursol yellow brown a, rodol yba, 3-nitro-6-aminophenol, 3-hydroxy-4-aminonitrobenzene, 2-amino-5-nitro-phenol, ccris 754, 4-amino-3-hydroxynitrobenzene | |
DOPJTDJKZNWLRB-UHFFFAOYSA-N | |
2-amino-5-nitrophenol | |
4984721 | |
154.13 | |
2-Amino-5-nitrophenol |
∼200°C (decomposition) | |
MFCD00007692 | |
972974 | |
Insoluble in water. Solubility in methanol gives very faint turbidity. Soluble in alcohol,benzene,acetone,and diethyl ether. | |
C1=CC(=C(C=C1[N+](=O)[O-])O)N | |
154.125 | |
CHEBI:82384 | |
96% |
Safety and Handling
GHS H Statement
H341-H315-H319-H335
Suspected of causing genetic defects.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P272-P280-P301+P312-P302+P352-P305+P351+P338-P330-P333+P313-P363-P501c
H302-H317-H319
EINECSNumber : 204-503-8
RTECSNumber : SJ6302500
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only