missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Amino-5-nitrothiazole, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 158520250

 View more versions of this product

Catalog No. AC158520250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

2-Amino-5-nitrothiazole
96.0
Authentic
Plastic bottle
MFCD00005326
15, 453
Solubility in water: slightly soluble. Other solubilities: soluble in dilute mineral acids,1g/100g alcohol,1g/250g ether,practically insoluble in chloroform
NC1=NC=C(S1)[N+]([O-])=O
145.14
CHEBI:82386
97%
121-66-4
100.0
96% min. (HPLC)
C3H3N3O2S
25g
2-amino-5-nitrothiazole, 5-nitrothiazol-2-amine, enheptin, entramin, 2-thiazolamine, 5-nitro, nitramin ido, amnizol soluble, enheptin-t, 5-nitro-2-thiazolamine, enheptin premix
MIHADVKEHAFNPG-UHFFFAOYSA-N
5-nitro-1,3-thiazol-2-amine
8486
145.14

Chemical Identifiers

121-66-4
145.14
MIHADVKEHAFNPG-UHFFFAOYSA-N
8486
5-nitro-1,3-thiazol-2-amine
C3H3N3O2S
MFCD00005326
2-amino-5-nitrothiazole, 5-nitrothiazol-2-amine, enheptin, entramin, 2-thiazolamine, 5-nitro, nitramin ido, amnizol soluble, enheptin-t, 5-nitro-2-thiazolamine, enheptin premix
CHEBI:82386
NC1=NC=C(S1)[N+]([O-])=O
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful in contact with skin.
Suspected of causing cancer.
Harmful if swallowed.
Harmful if inhaled.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Use personal protective equipment as required.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 204-490-9

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only