Learn More
2-Amino-5-nitrothiazole, 97%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 158521000
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
2-Amino-5-nitrothiazole | |
96.0 | |
Authentic | |
Plastic bottle | |
MFCD00005326 | |
15, 453 | |
Solubility in water: slightly soluble. Other solubilities: soluble in dilute mineral acids,1g/100g alcohol,1g/250g ether,practically insoluble in chloroform | |
NC1=NC=C(S1)[N+]([O-])=O | |
145.14 | |
CHEBI:82386 | |
97% |
121-66-4 | |
100.0 | |
96% min. (HPLC) | |
C3H3N3O2S | |
100g | |
2-amino-5-nitrothiazole, 5-nitrothiazol-2-amine, enheptin, entramin, 2-thiazolamine, 5-nitro, nitramin ido, amnizol soluble, enheptin-t, 5-nitro-2-thiazolamine, enheptin premix | |
MIHADVKEHAFNPG-UHFFFAOYSA-N | |
5-nitro-1,3-thiazol-2-amine | |
8486 | |
145.14 |
Chemical Identifiers
121-66-4 | |
145.14 | |
MIHADVKEHAFNPG-UHFFFAOYSA-N | |
8486 | |
5-nitro-1,3-thiazol-2-amine |
C3H3N3O2S | |
MFCD00005326 | |
2-amino-5-nitrothiazole, 5-nitrothiazol-2-amine, enheptin, entramin, 2-thiazolamine, 5-nitro, nitramin ido, amnizol soluble, enheptin-t, 5-nitro-2-thiazolamine, enheptin premix | |
CHEBI:82386 | |
NC1=NC=C(S1)[N+]([O-])=O |
Safety and Handling
GHS H Statement:
Harmful in contact with skin.
Suspected of causing cancer.
Harmful if swallowed.
Harmful if inhaled.
GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Use personal protective equipment as required.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
EINECSNumber : 204-490-9
RUO â Research Use Only