Learn More
2-Amino-5-phenylpyridine, 97%, Thermo Scientific Chemicals
$112.32 - $295.84
Chemical Identifiers
CAS | 33421-40-8 |
---|---|
Molecular Formula | C11H10N2 |
Molecular Weight (g/mol) | 170.22 |
MDL Number | MFCD01692452 |
InChI Key | OAPVIBHQRYFYSE-UHFFFAOYSA-N |
Synonym | 2-amino-5-phenylpyridine, 5-phenyl-2-pyridinamine, 2-pyridinamine, 5-phenyl, phe-p-1, 5-phenyl-pyridin-2-ylamine, 5-phenylpyridin-2-ylamine, 2-amino-5-phenyl pyridine, pyridine, 2-amino-5-phenyl, acmc-1adxr, 5-phenyl-2-pyridylamine |
PubChem CID | 105097 |
IUPAC Name | 5-phenylpyridin-2-amine |
SMILES | NC1=CC=C(C=N1)C1=CC=CC=C1 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
AAH26002MD
|
Thermo Scientific Chemicals
H26002MD |
250 mg |
N/A
|
|
||||||
AAH2600203
|
Thermo Scientific Chemicals
H2600203 |
1 g |
N/A
|
|
||||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
33421-40-8 | |
170.22 | |
OAPVIBHQRYFYSE-UHFFFAOYSA-N | |
105097 | |
NC1=CC=C(C=N1)C1=CC=CC=C1 |
C11H10N2 | |
MFCD01692452 | |
2-amino-5-phenylpyridine, 5-phenyl-2-pyridinamine, 2-pyridinamine, 5-phenyl, phe-p-1, 5-phenyl-pyridin-2-ylamine, 5-phenylpyridin-2-ylamine, 2-amino-5-phenyl pyridine, pyridine, 2-amino-5-phenyl, acmc-1adxr, 5-phenyl-2-pyridylamine | |
5-phenylpyridin-2-amine |
Specifications
33421-40-8 | |
C11H10N2 | |
250 mg | |
OAPVIBHQRYFYSE-UHFFFAOYSA-N | |
5-phenylpyridin-2-amine | |
105097 | |
97% |
133°C to 137°C | |
MFCD01692452 | |
2-amino-5-phenylpyridine, 5-phenyl-2-pyridinamine, 2-pyridinamine, 5-phenyl, phe-p-1, 5-phenyl-pyridin-2-ylamine, 5-phenylpyridin-2-ylamine, 2-amino-5-phenyl pyridine, pyridine, 2-amino-5-phenyl, acmc-1adxr, 5-phenyl-2-pyridylamine | |
NC1=CC=C(C=N1)C1=CC=CC=C1 | |
170.22 | |
170.21 | |
2-Amino-5-phenylpyridine |
Safety and Handling
GHS H Statement
H351-H315-H319-H335
Suspected of causing cancer.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P201-P202-P261-P264b-P271-P280i-P281-P302+P352-P304+P340-P305+P351+P338-P308+P313-P332+P313-P362-P501c
H315-H319-H335-H351
RTECSNumber : US2145000
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only