missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Amino-6-bromobenzothiazole, 95%, Thermo Scientific™

Chemical Identifiers

CAS 15864-32-1
Molecular Formula C7H5BrN2S
Molecular Weight (g/mol) 229.10
MDL Number MFCD00152229
InChI Key VZEBSJIOUMDNLY-UHFFFAOYSA-N
Synonym 2-amino-6-bromobenzothiazole, 6-bromobenzo d thiazol-2-amine, 2-benzothiazolamine, 6-bromo, 6-bromo-benzothiazol-2-ylamine, 6-bromobenzothiazol-2-ylamine, 6-bromo-2-aminobenzothiazole, benzothiazole, 2-amino-6-bromo, 6-bromo-2-benzothiazolamine, 2-amino-6-bromo benzothiazole, zerenex e/9072304
PubChem CID 85149
IUPAC Name 6-bromo-1,3-benzothiazol-2-amine
SMILES NC1=NC2=CC=C(Br)C=C2S1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC442290050
SDS View Documents Promotion Details
Thermo Scientific Chemicals
442290050
5g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC442290250
SDS View Documents Promotion Details
Thermo Scientific Chemicals
442290250
25g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

15864-32-1
229.10
VZEBSJIOUMDNLY-UHFFFAOYSA-N
85149
NC1=NC2=CC=C(Br)C=C2S1
C7H5BrN2S
MFCD00152229
2-amino-6-bromobenzothiazole, 6-bromobenzo d thiazol-2-amine, 2-benzothiazolamine, 6-bromo, 6-bromo-benzothiazol-2-ylamine, 6-bromobenzothiazol-2-ylamine, 6-bromo-2-aminobenzothiazole, benzothiazole, 2-amino-6-bromo, 6-bromo-2-benzothiazolamine, 2-amino-6-bromo benzothiazole, zerenex e/9072304
6-bromo-1,3-benzothiazol-2-amine

Specifications

15864-32-1
100.0
94% min. (HPLC)
C7H5BrN2S
5g
VZEBSJIOUMDNLY-UHFFFAOYSA-N
6-bromo-1,3-benzothiazol-2-amine
85149
95%
94.0
Authentic
Glass bottle
MFCD00152229
2-amino-6-bromobenzothiazole, 6-bromobenzo d thiazol-2-amine, 2-benzothiazolamine, 6-bromo, 6-bromo-benzothiazol-2-ylamine, 6-bromobenzothiazol-2-ylamine, 6-bromo-2-aminobenzothiazole, benzothiazole, 2-amino-6-bromo, 6-bromo-2-benzothiazolamine, 2-amino-6-bromo benzothiazole, zerenex e/9072304
NC1=NC2=CC=C(Br)C=C2S1
229.10
229.1
2-Amino-6-bromobenzothiazole
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
May cause an allergic skin reaction.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Contaminated work clothing should not be allowed out of the workplace.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only