missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Amino-6-fluorophenol, 97%, Thermo Scientific Chemicals

$168.62 - $682.64

Chemical Identifiers

CAS 53981-25-2
Molecular Formula C6H6FNO
Molecular Weight (g/mol) 127.118
MDL Number MFCD08532465
InChI Key MDIAVSZFIQWYST-UHFFFAOYSA-N
Synonym 6-fluoro-2-aminophenol, 3-fluoro-2-hydroxyaniline, 2-amino-6-fluoro-phenol, phenol, 2-amino-6-fluoro, pubchem10659, acmc-209lbm, 2-amino-6-flourophenol, phenol,2-amino-6-fluoro
PubChem CID 643562
IUPAC Name 2-amino-6-fluorophenol
SMILES C1=CC(=C(C(=C1)F)O)N
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6662603
View Documents
Thermo Scientific Chemicals
H6662603
1 g
Each for $168.62
 
AAH6662606
View Documents
Thermo Scientific Chemicals
H6662606
5 g
Each for $682.64
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

53981-25-2
127.118
MDIAVSZFIQWYST-UHFFFAOYSA-N
643562
C1=CC(=C(C(=C1)F)O)N
C6H6FNO
MFCD08532465
6-fluoro-2-aminophenol, 3-fluoro-2-hydroxyaniline, 2-amino-6-fluoro-phenol, phenol, 2-amino-6-fluoro, pubchem10659, acmc-209lbm, 2-amino-6-flourophenol, phenol,2-amino-6-fluoro
2-amino-6-fluorophenol

Specifications

53981-25-2
MFCD08532465
UN2811
6-fluoro-2-aminophenol, 3-fluoro-2-hydroxyaniline, 2-amino-6-fluoro-phenol, phenol, 2-amino-6-fluoro, pubchem10659, acmc-209lbm, 2-amino-6-flourophenol, phenol,2-amino-6-fluoro
C1=CC(=C(C(=C1)F)O)N
127.118
127.12
Solid
C6H6FNO
1 g
3603077
MDIAVSZFIQWYST-UHFFFAOYSA-N
2-amino-6-fluorophenol
643562
97%
2-Amino-6-fluorophenol
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302+H312+H332-H315-H319-H335

DOTInformation : Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

TSCA : No

storageNote1 : -30° to -10°C

Recommended Storage : Store at -20°C

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only