Learn More
2-Amino-6-methylbenzoic acid, 98%, Thermo Scientific Chemicals
$158.70 - $1455.47
Chemical Identifiers
CAS | 4389-50-8 |
---|---|
Molecular Formula | C8H8NO2 |
Molecular Weight (g/mol) | 150.16 |
MDL Number | MFCD00007809 |
InChI Key | XHYVBIXKORFHFM-UHFFFAOYSA-M |
Synonym | 6-methylanthranilic acid, 6-amino-o-toluic acid, benzoic acid, 2-amino-6-methyl, 2-amino-6-methyl-benzoic acid, 6-methylanthranilate, pubchem4957, acmc-209jvt, 6-methyl anthranilic acid, 3-amino-2-carboxytoluene, 2-carboxy-3-methylaniline |
PubChem CID | 151210 |
IUPAC Name | 2-amino-6-methylbenzoic acid |
SMILES | CC1=CC=CC(N)=C1C([O-])=O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
AAA1518706
|
Thermo Scientific Chemicals
A1518706 |
5 g |
Each for $158.70
|
|
||||||
AAA1518714
|
Thermo Scientific Chemicals
A1518714 |
25 g |
Each for $522.29
|
|
||||||
AAA1518722
|
Thermo Scientific Chemicals
A1518722 |
100 g |
Each for $1,455.47
|
|
||||||
Description
2-Amino-6-methylbenzoic acid reacts with cyanogen bromide to prepare 2-amino-5-methyl-benzo[d][1,3]oxazin-4-one.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2-Amino-6-methylbenzoic acid reacts with cyanogen bromide to prepare 2-amino-5-methyl-benzo[d][1,3]oxazin-4-one.
Solubility
Soluble in methanol.
Notes
Incompatible with strong oxidizing agents and strong acids.
Chemical Identifiers
4389-50-8 | |
150.16 | |
XHYVBIXKORFHFM-UHFFFAOYSA-M | |
151210 | |
CC1=CC=CC(N)=C1C([O-])=O |
C8H8NO2 | |
MFCD00007809 | |
6-methylanthranilic acid, 6-amino-o-toluic acid, benzoic acid, 2-amino-6-methyl, 2-amino-6-methyl-benzoic acid, 6-methylanthranilate, pubchem4957, acmc-209jvt, 6-methyl anthranilic acid, 3-amino-2-carboxytoluene, 2-carboxy-3-methylaniline | |
2-amino-6-methylbenzoic acid |
Specifications
4389-50-8 | |
C8H8NO2 | |
5 g | |
6-methylanthranilic acid, 6-amino-o-toluic acid, benzoic acid, 2-amino-6-methyl, 2-amino-6-methyl-benzoic acid, 6-methylanthranilate, pubchem4957, acmc-209jvt, 6-methyl anthranilic acid, 3-amino-2-carboxytoluene, 2-carboxy-3-methylaniline | |
XHYVBIXKORFHFM-UHFFFAOYSA-M | |
2-amino-6-methylbenzoic acid | |
151210 | |
98% |
∼127°C (decomposition) | |
MFCD00007809 | |
2803391 | |
Soluble in methanol. | |
CC1=CC=CC(N)=C1C([O-])=O | |
150.16 | |
151.16 | |
2-Amino-6-methylbenzoic acid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 000-000-0
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only