missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Amino-9,9-dimethylfluorene 99.0+%, TCI America™

$38.06 - $110.73

Chemical Identifiers

CAS 108714-73-4
Molecular Formula C15H15N
Molecular Weight (g/mol) 209.292
MDL Number MFCD09953790
InChI Key GUTJITRKAMCHSD-UHFFFAOYSA-N
Synonym 2-amino-9,9-dimethylfluorene, 9,9-dimethyl-9h-fluoren-2-amine, 2-amino-9,9-dimethyl-9h-fluorene, 9,9-dimethyl-9h-fluoren-2-ylamine, 9,9-dimethyl9h-fluorene-2-amine, pubchem23234, ksc495c6d, 9,9-dimethyl-2-aminofluorene, 2-amino-9,9-dimethyl fluorene, 9,9-dimethyl-fluoren-2-amine
PubChem CID 22617093
IUPAC Name 9,9-dimethylfluoren-2-amine
SMILES CC1(C2=CC=CC=C2C3=C1C=C(C=C3)N)C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
A2634200MG
View Documents
TCI America
A2634200MG
200 mg
Each for $38.06
 
A26341G
View Documents
TCI America
A26341G
1 g
Each for $110.73
 
Description

Description

Specifications

Chemical Identifiers

108714-73-4
209.292
GUTJITRKAMCHSD-UHFFFAOYSA-N
22617093
CC1(C2=CC=CC=C2C3=C1C=C(C=C3)N)C
C15H15N
MFCD09953790
2-amino-9,9-dimethylfluorene, 9,9-dimethyl-9h-fluoren-2-amine, 2-amino-9,9-dimethyl-9h-fluorene, 9,9-dimethyl-9h-fluoren-2-ylamine, 9,9-dimethyl9h-fluorene-2-amine, pubchem23234, ksc495c6d, 9,9-dimethyl-2-aminofluorene, 2-amino-9,9-dimethyl fluorene, 9,9-dimethyl-fluoren-2-amine
9,9-dimethylfluoren-2-amine

Specifications

108714-73-4
C15H15N
200 mg
GUTJITRKAMCHSD-UHFFFAOYSA-N
9,9-dimethylfluoren-2-amine
22617093
≥99.0% (HPLC,T)
2-Amino-9,9-dimethylfluorene
168°C
MFCD09953790
2-amino-9,9-dimethylfluorene, 9,9-dimethyl-9h-fluoren-2-amine, 2-amino-9,9-dimethyl-9h-fluorene, 9,9-dimethyl-9h-fluoren-2-ylamine, 9,9-dimethyl9h-fluorene-2-amine, pubchem23234, ksc495c6d, 9,9-dimethyl-2-aminofluorene, 2-amino-9,9-dimethyl fluorene, 9,9-dimethyl-fluoren-2-amine
CC1(C2=CC=CC=C2C3=C1C=C(C=C3)N)C
209.292
209.29
Crystalline Powder
Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
missing translation for 'documents'

missing translation for 'documents'