missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2-Amino-9,9-diphenylfluorene 98.0+%, TCI America™
Supplier: TCI America A27795G
Specifications
2-Amino-9,9-diphenylfluorene | |
Yellow | |
MFCD22376664 | |
MQRGCMXCVJPWHI-UHFFFAOYSA-N | |
9,9-diphenylfluoren-2-amine | |
70700696 | |
≥98.0% (HPLC,T) |
1268519-74-9 | |
C25H19N | |
5 g | |
C1=CC=C(C=C1)C2(C3=CC=CC=C3C4=C2C=C(C=C4)N)C5=CC=CC=C5 | |
333.434 | |
333.43 | |
Crystalline Powder |
Chemical Identifiers
1268519-74-9 | |
333.434 | |
MQRGCMXCVJPWHI-UHFFFAOYSA-N | |
9,9-diphenylfluoren-2-amine |
C25H19N | |
MFCD22376664 | |
70700696 | |
C1=CC=C(C=C1)C2(C3=CC=CC=C3C4=C2C=C(C=C4)N)C5=CC=CC=C5 |
Safety and Handling
TSCA : No