missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Amino-N-[(1,1'-biphenyl)-4-yl]-N-(4-bromophenyl)-9,9-dimethylfluorene 98.0+%, TCI America™
$59.65 - $268.43
Chemical Identifiers
| CAS | 1246562-40-2 |
|---|---|
| Molecular Formula | C33H26BrN |
| Molecular Weight (g/mol) | 516.48 |
| MDL Number | MFCD20040455 |
| InChI Key | CBLKFNHDNANUNU-UHFFFAOYSA-N |
| PubChem CID | 58131689 |
| IUPAC Name | N-{[1,1'-biphenyl]-4-yl}-N-(4-bromophenyl)-9,9-dimethyl-9H-fluoren-2-amine |
| SMILES | CC1(C)C2=CC=CC=C2C2=C1C=C(C=C2)N(C1=CC=C(Br)C=C1)C1=CC=C(C=C1)C1=CC=CC=C1 |
Chemical Identifiers
| 1246562-40-2 | |
| 516.48 | |
| CBLKFNHDNANUNU-UHFFFAOYSA-N | |
| N-{[1,1'-biphenyl]-4-yl}-N-(4-bromophenyl)-9,9-dimethyl-9H-fluoren-2-amine |
| C33H26BrN | |
| MFCD20040455 | |
| 58131689 | |
| CC1(C)C2=CC=CC=C2C2=C1C=C(C=C2)N(C1=CC=C(Br)C=C1)C1=CC=C(C=C1)C1=CC=CC=C1 |
Specifications
| 1246562-40-2 | |
| White-Yellow | |
| MFCD20040455 | |
| CBLKFNHDNANUNU-UHFFFAOYSA-N | |
| N-{[1,1'-biphenyl]-4-yl}-N-(4-bromophenyl)-9,9-dimethyl-9H-fluoren-2-amine | |
| 58131689 | |
| ≥98.0% (HPLC,N) | |
| 2-Amino-N-[(1,1′-biphenyl)-4-yl]-N-(4-bromophenyl)-9,9-dimethylfluorene |
| 161°C | |
| C33H26BrN | |
| 1 g | |
| CC1(C)C2=CC=CC=C2C2=C1C=C(C=C2)N(C1=CC=C(Br)C=C1)C1=CC=C(C=C1)C1=CC=CC=C1 | |
| 516.48 | |
| 516.48 | |
| Crystalline Powder |
Safety and Handling
TSCA : No