missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Amino-N-methylbenzamide 98.0+%, TCI America™
$108.11 - $400.78
Chemical Identifiers
| CAS | 4141-08-6 |
|---|---|
| Molecular Formula | C8H10N2O |
| Molecular Weight (g/mol) | 150.18 |
| MDL Number | MFCD00060602 |
| InChI Key | KIMWOULVHFLJIU-UHFFFAOYSA-N |
| Synonym | 2-Aminobenzoylmethylamide |
| PubChem CID | 308072 |
| ChEBI | CHEBI:81147 |
| IUPAC Name | 2-amino-N-methylbenzamide |
| SMILES | CNC(=O)C1=CC=CC=C1N |
Chemical Identifiers
| 4141-08-6 | |
| 150.18 | |
| KIMWOULVHFLJIU-UHFFFAOYSA-N | |
| 308072 | |
| 2-amino-N-methylbenzamide |
| C8H10N2O | |
| MFCD00060602 | |
| 2-Aminobenzoylmethylamide | |
| CHEBI:81147 | |
| CNC(=O)C1=CC=CC=C1N |
Specifications
| 4141-08-6 | |
| C8H10N2O | |
| 1 g | |
| KIMWOULVHFLJIU-UHFFFAOYSA-N | |
| 2-amino-N-methylbenzamide | |
| 308072 | |
| 150.18 | |
| Crystalline Powder |
| 79°C | |
| MFCD00060602 | |
| 2-Aminobenzoylmethylamide | |
| CNC(=O)C1=CC=CC=C1N | |
| 150.18 | |
| CHEBI:81147 | |
| ≥98.0% (GC,T) | |
| 2-Amino-N-methylbenzamide |
Safety and Handling
TSCA : No