missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2'-Aminoacetanilide, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals H2734314

 View more versions of this product

Catalog No. AAH2734314


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

2'-Aminoacetanilide
133°C to 137°C
MFCD00210388
n-2-aminophenyl acetamide, 2'-aminoacetanilide, o-aminoacetanilide, acetamide, n-2-aminophenyl, unii-lb34xrq95v, lb34xrq95v, acetamide, n-2-aminophenyl-9ci, n-acetyl-2-aminoaniline, n-acetyl-o-phenylenediamine, 2-aminoactanilide
CC(=O)NC1=CC=CC=C1N
150.18
150.18
34801-09-7
C8H10N2O
25 g
MPXAYYWSDIKNTP-UHFFFAOYSA-N
N-(2-aminophenyl)acetamide
11149
98%

Chemical Identifiers

34801-09-7
150.18
MPXAYYWSDIKNTP-UHFFFAOYSA-N
11149
CC(=O)NC1=CC=CC=C1N
C8H10N2O
MFCD00210388
n-2-aminophenyl acetamide, 2'-aminoacetanilide, o-aminoacetanilide, acetamide, n-2-aminophenyl, unii-lb34xrq95v, lb34xrq95v, acetamide, n-2-aminophenyl-9ci, n-acetyl-2-aminoaniline, n-acetyl-o-phenylenediamine, 2-aminoactanilide
N-(2-aminophenyl)acetamide
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only