missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2'-Aminoacetophenone, TCI America™
Supplier: TCI America A02505G
Specifications
2′-Aminoacetophenone | |
Yellow | |
C8H9NO | |
5 g | |
GTDQGKWDWVUKTI-UHFFFAOYSA-N | |
1-(2-aminophenyl)ethan-1-one | |
11086 | |
Liquid |
551-93-9 | |
252°C | |
MFCD00007717 | |
2'-aminoacetophenone, 1-2-aminophenyl ethanone, o-aminoacetophenone, 2-acetylaniline, o-acetylaniline, o-aminoacetylbenzene, 1-acetyl-2-aminobenzene, 1-2-aminophenyl ethan-1-one, acetophenone, 2'-amino, ethanone, 1-2-aminophenyl | |
CC(=O)C1=CC=CC=C1N | |
135.17 | |
135.17 |
Chemical Identifiers
551-93-9 | |
135.17 | |
GTDQGKWDWVUKTI-UHFFFAOYSA-N | |
11086 | |
CC(=O)C1=CC=CC=C1N |
C8H9NO | |
MFCD00007717 | |
2'-aminoacetophenone, 1-2-aminophenyl ethanone, o-aminoacetophenone, 2-acetylaniline, o-acetylaniline, o-aminoacetylbenzene, 1-acetyl-2-aminobenzene, 1-2-aminophenyl ethan-1-one, acetophenone, 2'-amino, ethanone, 1-2-aminophenyl | |
1-(2-aminophenyl)ethan-1-one |
Safety and Handling
RTECSNumber : AM5775000
TSCA : Yes