missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Aminobenzamide, 98+%, Thermo Scientific Chemicals

$65.21 - $241.05

Chemical Identifiers

CAS 88-68-6
Molecular Formula C7H8N2O
Molecular Weight (g/mol) 136.154
MDL Number MFCD00007981
InChI Key PXBFMLJZNCDSMP-UHFFFAOYSA-N
Synonym anthranilamide, benzamide, 2-amino, o-aminobenzamide, 2-carbamoylaniline, aminobenzamide, anthranilimidic acid, benzamide, o-amino, anthranilic acid amide, anthranilamide van, o-aminobenzamide van
PubChem CID 6942
IUPAC Name 2-aminobenzamide
SMILES C1=CC=C(C(=C1)C(=O)N)N
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1475622
View Documents
Thermo Scientific Chemicals
A1475622
100 g
Each for $65.21
 
AAA1475636
View Documents
Thermo Scientific Chemicals
A1475636
500 g
Each for $241.05
 
Description

Description

2-Aminobenzamide is used as an acetyldehyde scavenger in PET bottles. It is also used as an intermediate for ag chemicals, dyes, pharmaceuticals, blowing agents, and fine organic chemicals. In lubricating oils for supersonic jet engines, inhibits copper and magnesium corrosion.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Aminobenzamide is used as an acetyldehyde scavenger in PET bottles. It is also used as an intermediate for ag chemicals, dyes, pharmaceuticals, blowing agents, and fine organic chemicals. In lubricating oils for supersonic jet engines, inhibits copper and magnesium corrosion.

Solubility
Soluble in hot water and alcohol. Slightly soluble in ether and benzene. Very soluble in ethyl acetate.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
Specifications

Chemical Identifiers

88-68-6
136.154
PXBFMLJZNCDSMP-UHFFFAOYSA-N
6942
C1=CC=C(C(=C1)C(=O)N)N
C7H8N2O
MFCD00007981
anthranilamide, benzamide, 2-amino, o-aminobenzamide, 2-carbamoylaniline, aminobenzamide, anthranilimidic acid, benzamide, o-amino, anthranilic acid amide, anthranilamide van, o-aminobenzamide van
2-aminobenzamide

Specifications

88-68-6
300°C
C7H8N2O
100 g
anthranilamide, benzamide, 2-amino, o-aminobenzamide, 2-carbamoylaniline, aminobenzamide, anthranilimidic acid, benzamide, o-amino, anthranilic acid amide, anthranilamide van, o-aminobenzamide van
PXBFMLJZNCDSMP-UHFFFAOYSA-N
2-aminobenzamide
6942
≥98%
110°C to 114°C
198°C (388°F)
MFCD00007981
508509
Soluble in hot water and alcohol. Slightly soluble in ether and benzene. Very soluble in ethyl acetate.
C1=CC=C(C(=C1)C(=O)N)N
136.154
136.15
2-Aminobenzamide
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P280-P305+P351+P338-P310

H318

EINECSNumber : 201-851-2

RTECSNumber : CU8993000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only