missing translation for 'onlineSavingsMsg'
Learn More

2-Aminobenzamide 98.0+%, TCI America™

$178.95 - $1120.30

Chemical Identifiers

CAS 88-68-6
Molecular Formula C7H8N2O
Molecular Weight (g/mol) 136.154
MDL Number MFCD00007981
InChI Key PXBFMLJZNCDSMP-UHFFFAOYSA-N
Synonym anthranilamide, benzamide, 2-amino, o-aminobenzamide, 2-carbamoylaniline, aminobenzamide, anthranilimidic acid, benzamide, o-amino, anthranilic acid amide, anthranilamide van, o-aminobenzamide van
PubChem CID 6942
IUPAC Name 2-aminobenzamide
SMILES C1=CC=C(C(=C1)C(=O)N)N
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
A026225G
View Documents
TCI America
A026225G
25 g
Each for $178.95
Only null left
 
A0262100G
View Documents
TCI America
A0262100G
100 g
Each for $391.44
Only null left
 
A0262500G
View Documents
TCI America
A0262500G
500 g
Each for $1,120.30
Only null left
 
Description

Description

Specifications

Chemical Identifiers

88-68-6
136.154
PXBFMLJZNCDSMP-UHFFFAOYSA-N
6942
C1=CC=C(C(=C1)C(=O)N)N
C7H8N2O
MFCD00007981
anthranilamide, benzamide, 2-amino, o-aminobenzamide, 2-carbamoylaniline, aminobenzamide, anthranilimidic acid, benzamide, o-amino, anthranilic acid amide, anthranilamide van, o-aminobenzamide van
2-aminobenzamide

Specifications

88-68-6
White
MFCD00007981
anthranilamide, benzamide, 2-amino, o-aminobenzamide, 2-carbamoylaniline, aminobenzamide, anthranilimidic acid, benzamide, o-amino, anthranilic acid amide, anthranilamide van, o-aminobenzamide van
C1=CC=C(C(=C1)C(=O)N)N
136.154
136.15
Crystalline Powder
111°C
C7H8N2O
25 g
PXBFMLJZNCDSMP-UHFFFAOYSA-N
2-aminobenzamide
6942
≥98.0% (HPLC,T)
2-Aminobenzamide
Videos
Safety and Handling

Safety and Handling

EINECSNumber : (3)-4561

RTECSNumber : CU8993000

TSCA : Yes

SDS
missing translation for 'documents'

missing translation for 'documents'