missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Aminobenzophenone, 98%, Thermo Scientific Chemicals

$76.88 - $920.40

Chemical Identifiers

CAS 2835-77-0
Molecular Formula C13H11NO
Molecular Weight (g/mol) 197.24
MDL Number MFCD00007713
InChI Key MAOBFOXLCJIFLV-UHFFFAOYSA-N
Synonym 2-aminobenzophenone, 2-benzoylaniline, o-benzoylaniline, o-aminobenzophenone, 2-aminophenyl phenyl methanone, methanone, 2-aminophenyl phenyl, benzophenone, 2-amino, 2-amino-phenyl-phenyl-methanone, 2-aminophenyl phenyl ketone, methanone, aminophenyl phenyl
PubChem CID 76080
IUPAC Name (2-aminophenyl)-phenylmethanone
SMILES NC1=CC=CC=C1C(=O)C1=CC=CC=C1
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1258009
View Documents
Thermo Scientific Chemicals
A1258009
10 g
Each for $76.88
 
AAA1258018
View Documents
Thermo Scientific Chemicals
A1258018
50 g
Each for $252.01
 
AAA1258030
View Documents
Thermo Scientific Chemicals
A1258030
250 g
Each for $920.40
 
Description

Description

It is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and dyestuffs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and dyestuffs.

Solubility
Insoluble in water. Soluble in DMSO, Methanol.

Notes
Store at room temperature. Ensure good ventilation.
Specifications

Chemical Identifiers

2835-77-0
197.24
MAOBFOXLCJIFLV-UHFFFAOYSA-N
76080
NC1=CC=CC=C1C(=O)C1=CC=CC=C1
C13H11NO
MFCD00007713
2-aminobenzophenone, 2-benzoylaniline, o-benzoylaniline, o-aminobenzophenone, 2-aminophenyl phenyl methanone, methanone, 2-aminophenyl phenyl, benzophenone, 2-amino, 2-amino-phenyl-phenyl-methanone, 2-aminophenyl phenyl ketone, methanone, aminophenyl phenyl
(2-aminophenyl)-phenylmethanone

Specifications

2835-77-0
280°C
C13H11NO
10 g
2-aminobenzophenone, 2-benzoylaniline, o-benzoylaniline, o-aminobenzophenone, 2-aminophenyl phenyl methanone, methanone, 2-aminophenyl phenyl, benzophenone, 2-amino, 2-amino-phenyl-phenyl-methanone, 2-aminophenyl phenyl ketone, methanone, aminophenyl phenyl
MAOBFOXLCJIFLV-UHFFFAOYSA-N
(2-aminophenyl)-phenylmethanone
76080
98%
105°C to 108°C
240°C (464°F)
MFCD00007713
743425
Insoluble in water. Soluble in DMSO,Methanol.
NC1=CC=CC=C1C(=O)C1=CC=CC=C1
197.24
197.24
2-Aminobenzophenone
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 220-613-9

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only