missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Aminoethanethiol hydrochloride, 98%, Thermo Scientific Chemicals

$50.14 - $376.31

Chemical Identifiers

CAS 156-57-0
Molecular Formula C2H8ClNS
Molecular Weight (g/mol) 113.60
MDL Number MFCD00012904
InChI Key OGMADIBCHLQMIP-UHFFFAOYSA-N
Synonym cysteamine hydrochloride, 2-aminoethanethiol hydrochloride, cysteamine hcl, cysteaminium chloride, 2-mercaptoethylamine hydrochloride, 2-mercaptoethylamine hcl, bekaptan, mercaptamine hydrochloride, merkamin hydrochloride
PubChem CID 9082
SMILES [H+].[Cl-].NCCS
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC153770050
SDS View Documents
Thermo Scientific Chemicals
153770050
5 g Glass bottle
Each for $50.14
 
AC153770250
SDS View Documents
Thermo Scientific Chemicals
153770250
25 g Glass bottle
Each for $128.63
 
AC153771000
SDS View Documents
Thermo Scientific Chemicals
153771000
100 g Glass bottle
Each for $376.31
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

156-57-0
113.60
OGMADIBCHLQMIP-UHFFFAOYSA-N
9082
C2H8ClNS
MFCD00012904
cysteamine hydrochloride, 2-aminoethanethiol hydrochloride, cysteamine hcl, cysteaminium chloride, 2-mercaptoethylamine hydrochloride, 2-mercaptoethylamine hcl, bekaptan, mercaptamine hydrochloride, merkamin hydrochloride
[H+].[Cl-].NCCS

Specifications

156-57-0
100.0
White
Authentic
Glass bottle
H2NCH2CH2SH·HCl
5 g
15, 2776
Solubility in water: 400g/L (20°C). Other solubilities: very soluble in alcohol
[H+].[Cl-].NCCS
113.60
113.61
Crystalline Powder or Pellets
97.5
66.0°C to 70.0°C
1% max. (50°C, 4 hrs, 100mm Hg)
98%
C2H8ClNS
MFCD00012904
04, 286
cysteamine hydrochloride, 2-aminoethanethiol hydrochloride, cysteamine hcl, cysteaminium chloride, 2-mercaptoethylamine hydrochloride, 2-mercaptoethylamine hcl, bekaptan, mercaptamine hydrochloride, merkamin hydrochloride
OGMADIBCHLQMIP-UHFFFAOYSA-N
2-aminoethanethiol;hydrochloride
9082
98%
2-Aminoethanethiol hydrochloride
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
Harmful if swallowed.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 205-858-1

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only