missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Aminoethyl methacrylate hydrochloride, 90%, stabilized, Thermo Scientific Chemicals

$166.96 - $166.96

Chemical Identifiers

CAS 2420-94-2
Molecular Formula C6H11NO2·HCl
Molecular Weight (g/mol) 165.62
MDL Number MFCD00078260
InChI Key XSHISXQEKIKSGC-UHFFFAOYSA-N
Synonym 2-aminoethyl methacrylate hydrochloride, 2-aminoethylmethacrylate hydrochloride, 2-aminoethyl 2-methylacrylate hydrochloride, methacrylic acid, 2-aminoethyl ester, hydrochloride, polymers, 2-aminoethyl 2-methylprop-2-enoate hydrochloride, 2-propenoic acid, 2-methyl-, 2-aminoethyl ester, hydrochloride 1:1, 2-propenoic acid, 2-methyl-, 2-aminoethyl ester, hydrochloride, aminoethylmethacrylate, acmc-209wgm, timtec-bb sbb003905
PubChem CID 75495
IUPAC Name 2-aminoethyl 2-methylprop-2-enoate;hydrochloride
SMILES CC(=C)C(=O)OCCN.Cl
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC357810050
SDS View Documents
Thermo Scientific Chemicals
357810050
5 g Glass bottle
Each for $166.96
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

2420-94-2
165.62
XSHISXQEKIKSGC-UHFFFAOYSA-N
75495
CC(=C)C(=O)OCCN.Cl
C6H11NO2·HCl
MFCD00078260
2-aminoethyl methacrylate hydrochloride, 2-aminoethylmethacrylate hydrochloride, 2-aminoethyl 2-methylacrylate hydrochloride, methacrylic acid, 2-aminoethyl ester, hydrochloride, polymers, 2-aminoethyl 2-methylprop-2-enoate hydrochloride, 2-propenoic acid, 2-methyl-, 2-aminoethyl ester, hydrochloride 1:1, 2-propenoic acid, 2-methyl-, 2-aminoethyl ester, hydrochloride, aminoethylmethacrylate, acmc-209wgm, timtec-bb sbb003905
2-aminoethyl 2-methylprop-2-enoate;hydrochloride

Specifications

2420-94-2
100.0
Brown to White
88% min. (Argentometry)
C6H11NO2·HCl
5 g
Solubility in water: soluble
CC(=C)C(=O)OCCN.Cl
165.62
75495
90%
2-Aminoethyl methacrylate hydrochloride, 90%
88.0
116.0°C to 118.0°C
Authentic
Glass bottle
MFCD00078260
2-aminoethyl methacrylate hydrochloride, 2-aminoethylmethacrylate hydrochloride, 2-aminoethyl 2-methylacrylate hydrochloride, methacrylic acid, 2-aminoethyl ester, hydrochloride, polymers, 2-aminoethyl 2-methylprop-2-enoate hydrochloride, 2-propenoic acid, 2-methyl-, 2-aminoethyl ester, hydrochloride 1:1, 2-propenoic acid, 2-methyl-, 2-aminoethyl ester, hydrochloride, aminoethylmethacrylate, acmc-209wgm, timtec-bb sbb003905
XSHISXQEKIKSGC-UHFFFAOYSA-N
2-aminoethyl 2-methylprop-2-enoate;hydrochloride
500 to 1000ppm phenothiazine
165.62
Crystalline Powder
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 219-343-4

Recommended Storage : Refrigerator +4°C

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only