missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(+/-)-2-Aminoheptane, 98+%, Thermo Scientific Chemicals

$98.25 - $277.58

Chemical Identifiers

CAS 123-82-0
Molecular Formula C7H17N
Molecular Weight (g/mol) 115.22
MDL Number MFCD00008101
InChI Key VSRBKQFNFZQRBM-UHFFFAOYNA-N
Synonym 2-aminoheptane, tuaminoheptane, 2-heptylamine, 2-heptanamine, heptamine, 1-methylhexylamine, heptin, tuamine, tuaminoheptan, rineptil
PubChem CID 5603
IUPAC Name heptan-2-amine
SMILES CCCCCC(C)N
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAB2286314
View Documents
Thermo Scientific Chemicals
B2286314
25 g
Each for $98.25
 
AAB2286322
View Documents
Thermo Scientific Chemicals
B2286322
100 g
Each for $277.58
 
Description

Description

(±)-2-Aminoheptane is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(+/-)-2-Aminoheptane is used as an organic chemical synthesis intermediate.

Solubility
Soluble in water ( 9 g/L at 20°C).

Notes
Air sensitive. Incompatible with oxidizing agents and carbon dioxide.
Specifications

Chemical Identifiers

123-82-0
115.22
VSRBKQFNFZQRBM-UHFFFAOYNA-N
5603
CCCCCC(C)N
C7H17N
MFCD00008101
2-aminoheptane, tuaminoheptane, 2-heptylamine, 2-heptanamine, heptamine, 1-methylhexylamine, heptin, tuamine, tuaminoheptan, rineptil
heptan-2-amine

Specifications

123-82-0
11.45
33°C (91°F)
C7H17N
MFCD00008101
UN2733
Air Sensitive
2-aminoheptane, tuaminoheptane, 2-heptylamine, 2-heptanamine, heptamine, 1-methylhexylamine, heptin, tuamine, tuaminoheptan, rineptil
VSRBKQFNFZQRBM-UHFFFAOYNA-N
heptan-2-amine
5603
≥98%
0.768
142°C to 144°C
Amine-like
1.419
25 g
635676
14,9802
Soluble in water ( 9g/L at 20°C).
CCCCCC(C)N
115.22
115.22
(+/-)-2-Aminoheptane
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318-H226-H302-H312-H332
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapour.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.

P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c

H226-H302+H312+H332-H314-H335

DOTInformation : Hazard Class: 8; Packaging Group: II

EINECSNumber : 204-655-5

RTECSNumber : MQ5425000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only