missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Aminoisonicotinic acid, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 437360010

 View more versions of this product

Catalog No. AC437360010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

2-Aminoisonicotinic acid
96.0
Authentic
Glass bottle
1g
QMKZZQPPJRWDED-UHFFFAOYSA-N
2-aminopyridine-4-carboxylic acid
278396
97%
13362-28-2
100.0
96% min. (HPLC)
C6H6N2O2
2-aminoisonicotinic acid, 2-amino-4-pyridinecarboxylic acid, 2-amino-4-pyridine carboxylic acid, 2-amino-isonicotinic acid, 4-pyridinecarboxylic acid, 2-amino, 2-amino isonicotinic acid, pubchem2426, 2-aminoisonicotinicacid, acmc-209bt5, ksc174m9j
C1=CN=C(C=C1C(=O)O)N
138.13
138.13

Chemical Identifiers

13362-28-2
138.13
2-aminoisonicotinic acid, 2-amino-4-pyridinecarboxylic acid, 2-amino-4-pyridine carboxylic acid, 2-amino-isonicotinic acid, 4-pyridinecarboxylic acid, 2-amino, 2-amino isonicotinic acid, pubchem2426, 2-aminoisonicotinicacid, acmc-209bt5, ksc174m9j
2-aminopyridine-4-carboxylic acid
C6H6N2O2
QMKZZQPPJRWDED-UHFFFAOYSA-N
278396
C1=CN=C(C=C1C(=O)O)N
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only