missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-(Aminomethyl)pyridine, 98+%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals B2320822

 View more versions of this product

Catalog No. AAB2320822


Explore available promotions

Description

Description

2-(Amino methyl)pyridine is a synthesis reagent used in the synthesis of PPD analogues. It is also used in the preparation of 5-nitro-2-furancarboxylamides displaying potent tryoanocidal activity against trypanosomiasis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-(Amino methyl)pyridine is a synthesis reagent used in the synthesis of PPD analogues. It is also used in the preparation of 5-nitro-2-furancarboxylamides displaying potent tryoanocidal activity against trypanosomiasis.

Solubility
Soluble in water (1000 mg/ml at 20°C), and ethanol.

Notes
Air sensitive. Store in a cool, dry conditions, in well sealed conditions. Store away from oxidizing agents, acids, air.
Specifications

Specifications

2-(Aminomethyl)pyridine
-20°C
11 to 12
90°C (194°F)
C6H8N2
MFCD00006360
UN2735
Air Sensitive
Soluble in water (1000mg/ml at 20°C),and ethanol.
NCC1=CC=CC=N1
108.14
CHEBI:81387
99%
3731-51-9
1.049
202°C to 204°C
Amine-like
1.545
100 g
108054
2-picolylamine, 2-aminomethyl pyridine, 2-pyridinemethanamine, 2-aminomethylpyridine, 2-pyridinemethylamine, 2-pyridylmethylamine, 2-picolinamine, 2-pyridinemethaneamine, 2-pyridylmethyl amine, pyridine, 2-aminomethyl
WOXFMYVTSLAQMO-UHFFFAOYSA-N
pyridin-2-ylmethanamine
19509
108.14

Chemical Identifiers

3731-51-9
108.14
WOXFMYVTSLAQMO-UHFFFAOYSA-N
19509
NCC1=CC=CC=N1
C6H8N2
MFCD00006360
2-picolylamine, 2-aminomethyl pyridine, 2-pyridinemethanamine, 2-aminomethylpyridine, 2-pyridinemethylamine, 2-pyridylmethylamine, 2-picolinamine, 2-pyridinemethaneamine, 2-pyridylmethyl amine, pyridine, 2-aminomethyl
CHEBI:81387
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.

P210-P233-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P310-P312-P330-P331-P363-P374-P501c

H227-H302+H312-H314-H318-H335

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.

EINECSNumber : 223-090-5

RTECSNumber : US1840000

TSCA : Yes

Recommended Storage : Ambient temperatures; Store under Nitrogen

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only