missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Aminophenol, 99%, Thermo Scientific Chemicals

$62.45 - $661.27

Chemical Identifiers

CAS 95-55-6
Molecular Formula C6H7NO
Molecular Weight (g/mol) 109.128
MDL Number MFCD00007690
InChI Key CDAWCLOXVUBKRW-UHFFFAOYSA-N
Synonym o-aminophenol, o-hydroxyaniline, 2-hydroxyaniline, phenol, 2-amino, 2-amino-1-hydroxybenzene, fouramine op, benzofur gg, pelagol grey gg, pelagol 3ga, nako yellow 3ga
PubChem CID 5801
ChEBI CHEBI:18112
IUPAC Name 2-aminophenol
SMILES C1=CC=C(C(=C1)N)O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1373522
View Documents
Thermo Scientific Chemicals
A1373522
100 g
Each for $62.45
 
AAA1373536
View Documents
Thermo Scientific Chemicals
A1373536
500 g
Each for $190.23
 
AAA137350E
View Documents
Thermo Scientific Chemicals
A137350E
2500 g
Each for $661.27
 
Description

Description

2-Aminophenol is used to make dyes and pharmaceuticals. It is also used as a reagent for the synthesis of heterocyclic compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Aminophenol is used to make dyes and pharmaceuticals. It is also used as a reagent for the synthesis of heterocyclic compounds.

Solubility
Soluble in water (17 g/L at 20°C).

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
Specifications

Chemical Identifiers

95-55-6
109.128
CDAWCLOXVUBKRW-UHFFFAOYSA-N
5801
2-aminophenol
C6H7NO
MFCD00007690
o-aminophenol, o-hydroxyaniline, 2-hydroxyaniline, phenol, 2-amino, 2-amino-1-hydroxybenzene, fouramine op, benzofur gg, pelagol grey gg, pelagol 3ga, nako yellow 3ga
CHEBI:18112
C1=CC=C(C(=C1)N)O

Specifications

95-55-6
1.328
>175°C (347°F)
C6H7NO
100 g
606075
o-aminophenol, o-hydroxyaniline, 2-hydroxyaniline, phenol, 2-amino, 2-amino-1-hydroxybenzene, fouramine op, benzofur gg, pelagol grey gg, pelagol 3ga, nako yellow 3ga
CDAWCLOXVUBKRW-UHFFFAOYSA-N
2-aminophenol
5801
109.13
2-Aminophenol
173°C to 177°C
150°C (11 mmHg)
Characteristic
MFCD00007690
UN2512
14,461
Soluble in water (17g/L at 20°C).
C1=CC=C(C(=C1)N)O
109.128
CHEBI:18112
99%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H341-H302-H332
Suspected of causing genetic defects.
Harmful if swallowed.
Harmful if inhaled.

P201-P202-P261-P264b-P270-P271-P272-P280g-P281-P301+P312-P302+P352-P304+P340-P308+P313-P330-P333+P313-P363-P501c

H302+H332-H317-H341

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: AMINOPHENOLS (O-;M-;P-)

EINECSNumber : 202-431-1

RTECSNumber : SJ4950000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only