missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2-Aminopurine, 98%, Thermo Scientific Chemicals
A protein kinase R inhibitor
$335.82 - $954.48
Chemical Identifiers
CAS | 452-06-2 |
---|---|
Molecular Formula | C5H5N5 |
Molecular Weight (g/mol) | 135.13 |
MDL Number | MFCD00005566 |
InChI Key | MWBWWFOAEOYUST-UHFFFAOYSA-N |
Synonym | 2-aminopurine, 1h-purin-2-amine, 9h-purin-2-amine, purine, 2-amino, aminopurine, 2-amino purine, 2'-amino-purine, unii-o14b3u97fw, ccris 759, 9h-purin-2-ylamine |
PubChem CID | 9955 |
ChEBI | CHEBI:479072 |
IUPAC Name | 7H-purin-2-amine |
SMILES | NC1=NC=C2NC=NC2=N1 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAJ64919MD
|
Thermo Scientific Chemicals
J64919MD |
250 mg |
Each for $335.82
|
|
|||||
AAJ64919ME
|
Thermo Scientific Chemicals
J64919ME |
500 mg |
Each for $536.89
|
|
|||||
AAJ6491903
|
Thermo Scientific Chemicals
J6491903 |
1 g |
Each for $954.48
|
|
|||||
Description
A protein kinase R inhibitor
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
452-06-2 | |
135.13 | |
MWBWWFOAEOYUST-UHFFFAOYSA-N | |
9955 | |
7H-purin-2-amine |
C5H5N5 | |
MFCD00005566 | |
2-aminopurine, 1h-purin-2-amine, 9h-purin-2-amine, purine, 2-amino, aminopurine, 2-amino purine, 2'-amino-purine, unii-o14b3u97fw, ccris 759, 9h-purin-2-ylamine | |
CHEBI:479072 | |
NC1=NC=C2NC=NC2=N1 |
Specifications
452-06-2 | |
White to Yellow | |
C5H5N5 | |
250 mg | |
MWBWWFOAEOYUST-UHFFFAOYSA-N | |
7H-purin-2-amine | |
9955 | |
135.13 | |
Powder |
280°C to 282°C | |
Odorless | |
MFCD00005566 | |
2-aminopurine, 1h-purin-2-amine, 9h-purin-2-amine, purine, 2-amino, aminopurine, 2-amino purine, 2'-amino-purine, unii-o14b3u97fw, ccris 759, 9h-purin-2-ylamine | |
NC1=NC=C2NC=NC2=N1 | |
135.13 | |
CHEBI:479072 | |
98% | |
2-Aminopurine |
Safety and Handling
GHS H Statement
H302
Harmful if swallowed.
P264b-P270-P301+P312-P330-P501c
H302
EINECSNumber : 207-197-4
RTECSNumber : UO7475000
TSCA : No
Recommended Storage : Keep cold
RUO – Research Use Only