missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Aminopyrazine 98.0+%, TCI America™

$62.29 - $174.75

Chemical Identifiers

CAS 5049-61-6
Molecular Formula C4H5N3
Molecular Weight (g/mol) 95.105
MDL Number MFCD00006137
InChI Key XFTQRUTUGRCSGO-UHFFFAOYSA-N
Synonym 2-aminopyrazine, aminopyrazine, pyrazinamine, 2-pyrazinamine, pyrazin-2-ylamine, pyrazine-2-ylamine, pyrazine, 2-amino, amino pyrazine, 3-aminopyrazine, iminopyrazine
PubChem CID 78747
IUPAC Name pyrazin-2-amine
SMILES C1=CN=C(C=N1)N
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
A09895G
View Documents
TCI America
A09895G
5 g
Each for $62.29
 
A098925G
View Documents
TCI America
A098925G
25 g
Each for $174.75
 
Description

Description

Specifications

Chemical Identifiers

5049-61-6
95.105
XFTQRUTUGRCSGO-UHFFFAOYSA-N
78747
C1=CN=C(C=N1)N
C4H5N3
MFCD00006137
2-aminopyrazine, aminopyrazine, pyrazinamine, 2-pyrazinamine, pyrazin-2-ylamine, pyrazine-2-ylamine, pyrazine, 2-amino, amino pyrazine, 3-aminopyrazine, iminopyrazine
pyrazin-2-amine

Specifications

5049-61-6
C4H5N3
5 g
XFTQRUTUGRCSGO-UHFFFAOYSA-N
pyrazin-2-amine
78747
≥98.0% (GC,T)
2-Aminopyrazine
121°C
MFCD00006137
2-aminopyrazine, aminopyrazine, pyrazinamine, 2-pyrazinamine, pyrazin-2-ylamine, pyrazine-2-ylamine, pyrazine, 2-amino, amino pyrazine, 3-aminopyrazine, iminopyrazine
C1=CN=C(C=N1)N
95.105
95.11
Crystalline Powder
Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
Documents

Documents