missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2-Amyl-5-methylphenol 98.0+%, TCI America™
$218.00 - $655.71
Chemical Identifiers
CAS | 1300-94-3 |
---|---|
Molecular Formula | C12H18O |
Molecular Weight (g/mol) | 178.275 |
MDL Number | MFCD00127710 |
InChI Key | CKGWFZQGEQJZIL-UHFFFAOYSA-N |
Synonym | 5-Methyl-2-pentylphenol |
PubChem CID | 14759 |
ChEBI | CHEBI:48213 |
IUPAC Name | 5-methyl-2-pentylphenol |
SMILES | CCCCCC1=C(C=C(C=C1)C)O |
Chemical Identifiers
1300-94-3 | |
178.275 | |
CKGWFZQGEQJZIL-UHFFFAOYSA-N | |
14759 | |
5-methyl-2-pentylphenol |
C12H18O | |
MFCD00127710 | |
5-Methyl-2-pentylphenol | |
CHEBI:48213 | |
CCCCCC1=C(C=C(C=C1)C)O |
Specifications
1300-94-3 | |
C12H18O | |
1 g | |
5-Methyl-2-pentylphenol | |
CCCCCC1=C(C=C(C=C1)C)O | |
178.275 | |
CHEBI:48213 | |
≥98.0% (GC) | |
2-Amyl-5-methylphenol |
139°C | |
MFCD00127710 | |
3145 | |
CKGWFZQGEQJZIL-UHFFFAOYSA-N | |
5-methyl-2-pentylphenol | |
14759 | |
178.28 | |
Liquid |
Safety and Handling
EINECSNumber : (3)-0521
RTECSNumber : GP3100000
TSCA : No