missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
(-)-2-Azabicyclo[2.2.1]hept-5-en-3-one, 98%, > 95% ee, Thermo Scientific™
Chemical Identifiers
CAS | 79200-56-9 |
---|---|
Molecular Formula | C6H7NO |
Molecular Weight (g/mol) | 109.13 |
MDL Number | MFCD00211274 |
InChI Key | DDUFYKNOXPZZIW-UHNVWZDZSA-N |
Synonym | 1r,4s-2-azabicyclo 2.2.1 hept-5-en-3-one, --1r,4s-2-azabicyclo 2.2.1 hept-5-en-3-one, 1s,4r-3-azabicyclo 2.2.1 hept-5-en-2-one, 2-azabicyclo 2,2,1 hept-5-en-3-one, vince lactam, 2-azabicyclo 2.2.1 hept-5-en-3-one, 1r,4s, --2-azabicyclo 2.2.1 hept-5-en-3-one, --vince lactam, 1s-+-2-azabicyclo 2.2.1 hept-5-en-3-one |
PubChem CID | 2725037 |
IUPAC Name | (1S,4R)-3-azabicyclo[2.2.1]hept-5-en-2-one |
SMILES | C1C2C=CC1NC2=O |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC297040010
|
Thermo Scientific Chemicals
297040010 |
1g | Glass bottle |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
AC297040050
|
Thermo Scientific Chemicals
297040050 |
5g | Glass bottle |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
79200-56-9 | |
109.13 | |
DDUFYKNOXPZZIW-UHNVWZDZSA-N | |
2725037 | |
C1C2C=CC1NC2=O |
C6H7NO | |
MFCD00211274 | |
1r,4s-2-azabicyclo 2.2.1 hept-5-en-3-one, --1r,4s-2-azabicyclo 2.2.1 hept-5-en-3-one, 1s,4r-3-azabicyclo 2.2.1 hept-5-en-2-one, 2-azabicyclo 2,2,1 hept-5-en-3-one, vince lactam, 2-azabicyclo 2.2.1 hept-5-en-3-one, 1r,4s, --2-azabicyclo 2.2.1 hept-5-en-3-one, --vince lactam, 1s-+-2-azabicyclo 2.2.1 hept-5-en-3-one | |
(1S,4R)-3-azabicyclo[2.2.1]hept-5-en-2-one |
Specifications
79200-56-9 | |
(c=1, CH2Cl2) (1 cm cell), (c=1, CH2Cl2) (1 cm cell) at 0nm, 0.1 max. at 400nm | |
95% min. (HPLC) | |
97.5% min. (HPLC) | |
C6H7NO | |
1g | |
1r,4s-2-azabicyclo 2.2.1 hept-5-en-3-one, --1r,4s-2-azabicyclo 2.2.1 hept-5-en-3-one, 1s,4r-3-azabicyclo 2.2.1 hept-5-en-2-one, 2-azabicyclo 2,2,1 hept-5-en-3-one, vince lactam, 2-azabicyclo 2.2.1 hept-5-en-3-one, 1r,4s, --2-azabicyclo 2.2.1 hept-5-en-3-one, --vince lactam, 1s-+-2-azabicyclo 2.2.1 hept-5-en-3-one | |
C1C2C=CC1NC2=O | |
109.13 | |
109.13 | |
(-)-2-Azabicyclo[2.2.1]hept-5-en-3-one, >95%ee |
97.5 | |
100.0 | |
Authentic | |
Glass bottle | |
MFCD00211274 | |
-560 | |
DDUFYKNOXPZZIW-UHNVWZDZSA-N | |
(1S,4R)-3-azabicyclo[2.2.1]hept-5-en-2-one | |
2725037 | |
98%,>95% ee |
Safety and Handling
GHS H Statement
Causes serious eye damage.
May cause an allergic skin reaction.
Harmful if swallowed.
GHS P Statement
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective
GHS Signal Word: Danger
RUO â Research Use Only