missing translation for 'onlineSavingsMsg'
Learn More

2-Benzoxazolinone, 98%

CAS: 59-49-4 | C7H5NO2 | 135.12 g/mol

$95.68 - $255.97

Chemical Identifiers

CAS 59-49-4
Molecular Formula C7H5NO2
Molecular Weight (g/mol) 135.12
MDL Number MFCD00005716
InChI Key ASSKVPFEZFQQNQ-UHFFFAOYSA-N
Synonym 2-benzoxazolinone, benzoxazolinone, benzoxazolone, 2-hydroxybenzoxazole, 2-benzoxazolol, 2 3h-benzoxazolone, 1,3-benzoxazol-2 3h-one, 2-benzoxazolone, benzo d oxazol-2 3h-one, benzoxazolin-2-one
PubChem CID 6043
IUPAC Name 2,3-dihydro-1,3-benzoxazol-2-one
SMILES O=C1NC2=CC=CC=C2O1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1937814
View Documents
Thermo Scientific Chemicals
A1937814
25 g
Each for $95.68
Only null left
 
AAA1937822
View Documents
Thermo Scientific Chemicals
A1937822
100 g
Each for $255.97
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

59-49-4
135.12
ASSKVPFEZFQQNQ-UHFFFAOYSA-N
6043
O=C1NC2=CC=CC=C2O1
C7H5NO2
MFCD00005716
2-benzoxazolinone, benzoxazolinone, benzoxazolone, 2-hydroxybenzoxazole, 2-benzoxazolol, 2 3h-benzoxazolone, 1,3-benzoxazol-2 3h-one, 2-benzoxazolone, benzo d oxazol-2 3h-one, benzoxazolin-2-one
2,3-dihydro-1,3-benzoxazol-2-one

Specifications

59-49-4
160°C (320°F)
MFCD00005716
119481
ASSKVPFEZFQQNQ-UHFFFAOYSA-N
2,3-dihydro-1,3-benzoxazol-2-one
6043
98%
139°C to 140°C
C7H5NO2
25 g
2-benzoxazolinone, benzoxazolinone, benzoxazolone, 2-hydroxybenzoxazole, 2-benzoxazolol, 2 3h-benzoxazolone, 1,3-benzoxazol-2 3h-one, 2-benzoxazolone, benzo d oxazol-2 3h-one, benzoxazolin-2-one
O=C1NC2=CC=CC=C2O1
135.12
135.12
2-Benzoxazolinone
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H332
Toxic if swallowed.
Harmful if inhaled.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P501c

H302+H312+H332

DOTInformation : Hazard Class: 6.1; Packaging Group: III

EINECSNumber : 200-430-0

RTECSNumber : DM4905000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only