missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Benzoylthiophene, 98%
CAS: 135-00-2 | C11H8OS | 188.244 g/mol
$71.14 - $319.52
Chemical Identifiers
| CAS | 135-00-2 |
|---|---|
| Molecular Formula | C11H8OS |
| Molecular Weight (g/mol) | 188.244 |
| MDL Number | MFCD00022495 |
| InChI Key | DWYFUJJWTRPARQ-UHFFFAOYSA-N |
| Synonym | 2-benzoylthiophene, phenyl thiophen-2-yl methanone, phenyl 2-thienyl ketone, methanone, phenyl-2-thienyl, ketone, phenyl 2-thienyl, phenyl 2-thienyl methanone, 2-thienyl phenyl ketone, .alpha.-benzoylthiophene, phenyl-thiophen-2-yl-methanone, 2-benzoyl thiophene |
| PubChem CID | 67262 |
| IUPAC Name | phenyl(thiophen-2-yl)methanone |
| SMILES | C1=CC=C(C=C1)C(=O)C2=CC=CS2 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAA1203606
|
Thermo Scientific Chemicals
A1203606 |
5 g |
Each for $71.14
|
|
|||||
|
AAA1203609
|
Thermo Scientific Chemicals
A1203609 |
10 g |
Each for $94.08
|
|
|||||
|
AAA1203618
|
Thermo Scientific Chemicals
A1203618 |
50 g |
Each for $319.52
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 135-00-2 | |
| 188.244 | |
| DWYFUJJWTRPARQ-UHFFFAOYSA-N | |
| 67262 | |
| C1=CC=C(C=C1)C(=O)C2=CC=CS2 |
| C11H8OS | |
| MFCD00022495 | |
| 2-benzoylthiophene, phenyl thiophen-2-yl methanone, phenyl 2-thienyl ketone, methanone, phenyl-2-thienyl, ketone, phenyl 2-thienyl, phenyl 2-thienyl methanone, 2-thienyl phenyl ketone, .alpha.-benzoylthiophene, phenyl-thiophen-2-yl-methanone, 2-benzoyl thiophene | |
| phenyl(thiophen-2-yl)methanone |
Specifications
| 135-00-2 | |
| 124°C to 126°C (0.5 mmHg) | |
| MFCD00022495 | |
| 3929 | |
| DWYFUJJWTRPARQ-UHFFFAOYSA-N | |
| phenyl(thiophen-2-yl)methanone | |
| 67262 | |
| 98% |
| 55°C to 57°C | |
| C11H8OS | |
| 5 g | |
| 2-benzoylthiophene, phenyl thiophen-2-yl methanone, phenyl 2-thienyl ketone, methanone, phenyl-2-thienyl, ketone, phenyl 2-thienyl, phenyl 2-thienyl methanone, 2-thienyl phenyl ketone, .alpha.-benzoylthiophene, phenyl-thiophen-2-yl-methanone, 2-benzoyl thiophene | |
| C1=CC=C(C=C1)C(=O)C2=CC=CS2 | |
| 188.244 | |
| 188.25 | |
| 2-Benzoylthiophene |
Safety and Handling
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P501c
H302+H312+H332
DOTInformation : Hazard Class: 6.1; Packaging Group: III
EINECSNumber : 205-169-6
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only