Learn More
2-Benzyl-2-imidazoline hydrochloride, 99%, Thermo Scientific Chemicals
$64.14 - $64.14
Chemical Identifiers
CAS | 59-97-2 |
---|---|
Molecular Formula | C10H13ClN2 |
Molecular Weight (g/mol) | 196.678 |
MDL Number | MFCD00012693 |
InChI Key | RHTNTTODYGNRSP-UHFFFAOYSA-N |
Synonym | tolazoline hydrochloride, tolazoline hcl, 2-benzyl-4,5-dihydro-1h-imidazole hydrochloride, arterodyl, tolavad, tolpal, priscoline hydrochloride, tolazoline chloride, imidaline hydrochloride, benzazoline hydrochloride |
PubChem CID | 6048 |
IUPAC Name | 2-benzyl-4,5-dihydro-1H-imidazole;hydrochloride |
SMILES | C1CN=C(N1)CC2=CC=CC=C2.Cl |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAB2176414
|
Thermo Scientific Chemicals
B2176414 |
25 g |
Each for $64.14
|
|
|||||
Description
An alpha adrenergic blocking agent
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
59-97-2 | |
196.678 | |
RHTNTTODYGNRSP-UHFFFAOYSA-N | |
6048 | |
C1CN=C(N1)CC2=CC=CC=C2.Cl |
C10H13ClN2 | |
MFCD00012693 | |
tolazoline hydrochloride, tolazoline hcl, 2-benzyl-4,5-dihydro-1h-imidazole hydrochloride, arterodyl, tolavad, tolpal, priscoline hydrochloride, tolazoline chloride, imidaline hydrochloride, benzazoline hydrochloride | |
2-benzyl-4,5-dihydro-1H-imidazole;hydrochloride |
Specifications
59-97-2 | |
C10H13ClN2 | |
25 g | |
14,9506 | |
RHTNTTODYGNRSP-UHFFFAOYSA-N | |
2-benzyl-4,5-dihydro-1H-imidazole;hydrochloride | |
6048 | |
99% |
174°C to 176°C | |
MFCD00012693 | |
Hygroscopic | |
tolazoline hydrochloride, tolazoline hcl, 2-benzyl-4,5-dihydro-1h-imidazole hydrochloride, arterodyl, tolavad, tolpal, priscoline hydrochloride, tolazoline chloride, imidaline hydrochloride, benzazoline hydrochloride | |
C1CN=C(N1)CC2=CC=CC=C2.Cl | |
196.678 | |
196.68 | |
2-Benzyl-2-imidazoline hydrochloride |
Safety and Handling
GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319
EINECSNumber : 200-447-3
TSCA : Yes
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only