missing translation for 'onlineSavingsMsg'
Learn More

2-(BOC-Amino)ethanethiol, 97%

CAS: 67385-09-5 | C7H15NO2S | 177.26 g/mol

$152.50 - $510.21

Chemical Identifiers

CAS 67385-09-5
Molecular Formula C7H15NO2S
Molecular Weight (g/mol) 177.26
MDL Number MFCD00274335
InChI Key GSJJCZSHYJNRPN-UHFFFAOYSA-N
Synonym 2-boc-amino ethanethiol, boc-cysteamine, tert-butyl n-2-mercaptoethyl carbamate, tert-butyl n-2-sulfanylethyl carbamate, 2-bocamino ethanethiol, tert-butyl 2-mercaptoethyl carbamate, carbamic acid, 2-mercaptoethyl-, 1,1-dimethylethyl ester, n-boc-aet, tert-butoxy-n-2-sulfanylethyl carboxamide, n-boc-cysteamine
PubChem CID 3017761
IUPAC Name tert-butyl N-(2-sulfanylethyl)carbamate
SMILES CC(C)(C)OC(=O)NCCS
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC381080050
SDS View Documents
Thermo Scientific Chemicals
381080050
5 mL Glass Bottle
Each for $152.50
Only null left
 
AC381080250
SDS View Documents
Thermo Scientific Chemicals
381080250
25 mL Glass Bottle
Each for $510.21
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

67385-09-5
177.26
GSJJCZSHYJNRPN-UHFFFAOYSA-N
3017761
CC(C)(C)OC(=O)NCCS
C7H15NO2S
MFCD00274335
2-boc-amino ethanethiol, boc-cysteamine, tert-butyl n-2-mercaptoethyl carbamate, tert-butyl n-2-sulfanylethyl carbamate, 2-bocamino ethanethiol, tert-butyl 2-mercaptoethyl carbamate, carbamic acid, 2-mercaptoethyl-, 1,1-dimethylethyl ester, n-boc-aet, tert-butoxy-n-2-sulfanylethyl carboxamide, n-boc-cysteamine
tert-butyl N-(2-sulfanylethyl)carbamate

Specifications

67385-09-5
96.0
Colorless to Yellow
96% min. (GC)
Glass Bottle
(CH3)3COC(O)NH(CH2)2SH
2-boc-amino ethanethiol, boc-cysteamine, tert-butyl n-2-mercaptoethyl carbamate, tert-butyl n-2-sulfanylethyl carbamate, 2-bocamino ethanethiol, tert-butyl 2-mercaptoethyl carbamate, carbamic acid, 2-mercaptoethyl-, 1,1-dimethylethyl ester, n-boc-aet, tert-butoxy-n-2-sulfanylethyl carboxamide, n-boc-cysteamine
1.0490g/mL
tert-butyl N-(2-sulfanylethyl)carbamate
5 mL
3017761
97%
Low Melting Solid or Viscous Liquid
68.0°C (0.3 mmHg)
100.0
Authentic
C7H15NO2S
1.4730 to 1.4770
MFCD00274335
GSJJCZSHYJNRPN-UHFFFAOYSA-N
CC(C)(C)OC(=O)NCCS
177.26
109°C
177.27
1.049
2-(BOC-Amino)ethanethiol
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only