missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Bromo-1,3,5-trifluorobenzene, 98+%, Thermo Scientific Chemicals

 

$112.48 - $112.48

Chemical Identifiers

CAS 2367-76-2
Molecular Formula C6H2BrF3
Molecular Weight (g/mol) 210.98
MDL Number MFCD00000334
InChI Key PZBSPSOGEVCRQI-UHFFFAOYSA-N
Synonym 1-bromo-2,4,6-trifluorobenzene, 2,4,6-trifluorobromobenzene, 2-bromo-1,3,5-trifluoro-benzene, benzene, 2-bromo-1,3,5-trifluoro, 1-bromo-2,4,6-trifluoro benzene, 1,3,5-trifluoro-2-bromobenzene, fr cf ef be, pubchem4306, acmc-1cbzk, intermediates-zcf02629
PubChem CID 75398
IUPAC Name 2-bromo-1,3,5-trifluorobenzene
SMILES FC1=CC(F)=C(Br)C(F)=C1
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1992006
View Documents
Thermo Scientific Chemicals
A1992006
5 g
Each for $112.48
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

 
Specifications

Chemical Identifiers

2367-76-2
210.98
PZBSPSOGEVCRQI-UHFFFAOYSA-N
75398
FC1=CC(F)=C(Br)C(F)=C1
C6H2BrF3
MFCD00000334
1-bromo-2,4,6-trifluorobenzene, 2,4,6-trifluorobromobenzene, 2-bromo-1,3,5-trifluoro-benzene, benzene, 2-bromo-1,3,5-trifluoro, 1-bromo-2,4,6-trifluoro benzene, 1,3,5-trifluoro-2-bromobenzene, fr cf ef be, pubchem4306, acmc-1cbzk, intermediates-zcf02629
2-bromo-1,3,5-trifluorobenzene

Specifications

2367-76-2
1.79
58°C (136°F)
1.485
5 g
2502668
PZBSPSOGEVCRQI-UHFFFAOYSA-N
2-bromo-1,3,5-trifluorobenzene
75398
≥98%
3°C to 4°C
140°C to 141°C
C6H2BrF3
MFCD00000334
UN1993
1-bromo-2,4,6-trifluorobenzene, 2,4,6-trifluorobromobenzene, 2-bromo-1,3,5-trifluoro-benzene, benzene, 2-bromo-1,3,5-trifluoro, 1-bromo-2,4,6-trifluoro benzene, 1,3,5-trifluoro-2-bromobenzene, fr cf ef be, pubchem4306, acmc-1cbzk, intermediates-zcf02629
FC1=CC(F)=C(Br)C(F)=C1
210.98
210.98
2-Bromo-1,3,5-trifluorobenzene
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c

H226-H315-H319-H335

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.

EINECSNumber : 219-125-9

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only