missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2-Bromo-1,3,5-triisopropylbenzene, 96%, Thermo Scientific Chemicals
$188.53 - $2629.38
Chemical Identifiers
CAS | 21524-34-5 |
---|---|
Molecular Formula | C15H23Br |
Molecular Weight (g/mol) | 283.253 |
MDL Number | MFCD00051547 |
InChI Key | FUMMYHVKFAHQST-UHFFFAOYSA-N |
Synonym | 2-bromo-1,3,5-triisopropylbenzene, 1-bromo-2,4,6-triisopropylbenzene, 2,4,6-triisopropylbromobenzene, 1,3,5-triisopropyl-2-bromobenzene, 2-bromo-1,3,5-tris propan-2-yl benzene, 2-bromo-1,3,5-triisopropyl-benzene, 2-bromo-1,3,5-tri propan-2-yl benzene, benzene, 2-bromo-1,3,5-tris 1-methylethyl, 2-bromo-1,3,5-tris 1-methylethyl-benzene, acmc-209flf |
PubChem CID | 140846 |
IUPAC Name | 2-bromo-1,3,5-tri(propan-2-yl)benzene |
SMILES | CC(C)C1=CC(=C(C(=C1)C(C)C)Br)C(C)C |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAA1569214
|
Thermo Scientific Chemicals
A1569214 |
25 g |
Each for $188.53
|
|
|||||
AAA1569222
|
Thermo Scientific Chemicals
A1569222 |
100 g |
Each for $660.44
|
|
|||||
AAA1569236
|
Thermo Scientific Chemicals
A1569236 |
500 g |
Each for $2,629.38
|
|
|||||
Description
2-Bromo-1,3,5-triisopropylbenzene is used in Suzuki reaction. It is found to effect highly selective ring closures of homoallylic alcohols to substituted tetrahydrofurans.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2-Bromo-1,3,5-triisopropylbenzene is used in Suzuki reaction. It is found to effect highly selective ring closures of homoallylic alcohols to substituted tetrahydrofurans.
Solubility
Slightly miscible with water.
Notes
Incompatible with strong oxidizing agents.
Chemical Identifiers
21524-34-5 | |
283.253 | |
FUMMYHVKFAHQST-UHFFFAOYSA-N | |
140846 | |
CC(C)C1=CC(=C(C(=C1)C(C)C)Br)C(C)C |
C15H23Br | |
MFCD00051547 | |
2-bromo-1,3,5-triisopropylbenzene, 1-bromo-2,4,6-triisopropylbenzene, 2,4,6-triisopropylbromobenzene, 1,3,5-triisopropyl-2-bromobenzene, 2-bromo-1,3,5-tris propan-2-yl benzene, 2-bromo-1,3,5-triisopropyl-benzene, 2-bromo-1,3,5-tri propan-2-yl benzene, benzene, 2-bromo-1,3,5-tris 1-methylethyl, 2-bromo-1,3,5-tris 1-methylethyl-benzene, acmc-209flf | |
2-bromo-1,3,5-tri(propan-2-yl)benzene |
Specifications
21524-34-5 | |
C15H23Br | |
MFCD00051547 | |
1953440 | |
Slightly miscible with water. | |
CC(C)C1=CC(=C(C(=C1)C(C)C)Br)C(C)C | |
283.253 | |
283.25 | |
2-Bromo-1,3,5-triisopropylbenzene |
125°C to 126°C (4 mmHg) | |
1.5225 | |
25 g | |
2-bromo-1,3,5-triisopropylbenzene, 1-bromo-2,4,6-triisopropylbenzene, 2,4,6-triisopropylbromobenzene, 1,3,5-triisopropyl-2-bromobenzene, 2-bromo-1,3,5-tris propan-2-yl benzene, 2-bromo-1,3,5-triisopropyl-benzene, 2-bromo-1,3,5-tri propan-2-yl benzene, benzene, 2-bromo-1,3,5-tris 1-methylethyl, 2-bromo-1,3,5-tris 1-methylethyl-benzene, acmc-209flf | |
FUMMYHVKFAHQST-UHFFFAOYSA-N | |
2-bromo-1,3,5-tri(propan-2-yl)benzene | |
140846 | |
96% |
Safety and Handling
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only