missing translation for 'onlineSavingsMsg'
Learn More

2-Bromo-1,3,5-trimethyl-4-nitrobenzene, 90+%

CAS: 90561-85-6 | C9H10BrNO2 | 244.088 g/mol

$119.68 - $400.73

Chemical Identifiers

CAS 90561-85-6
Molecular Formula C9H10BrNO2
Molecular Weight (g/mol) 244.088
MDL Number MFCD00015924
InChI Key IUFDRRDWXUSFBG-UHFFFAOYSA-N
Synonym 3-bromo-2,4,6-trimethylnitrobenzene, 2-bromo-4-nitro-1,3,5-trimethylbenzene, 1-bromo-3-nitro-2,4,6-trimethylbenzene, 4-bromo-2-nitromesitylene, 4-bromo-1,3,5-trimethyl-2-nitrobenzene, 2-bromanyl-1,3,5-trimethyl-4-nitro-benzene
PubChem CID 2757200
IUPAC Name 2-bromo-1,3,5-trimethyl-4-nitrobenzene
SMILES CC1=CC(=C(C(=C1[N+](=O)[O-])C)Br)C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL0510703
View Documents
Thermo Scientific Chemicals
L0510703
1 g N/A
Only null left
 
AAL0510706
View Documents
Thermo Scientific Chemicals
L0510706
5 g N/A
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

90561-85-6
244.088
IUFDRRDWXUSFBG-UHFFFAOYSA-N
2757200
CC1=CC(=C(C(=C1[N+](=O)[O-])C)Br)C
C9H10BrNO2
MFCD00015924
3-bromo-2,4,6-trimethylnitrobenzene, 2-bromo-4-nitro-1,3,5-trimethylbenzene, 1-bromo-3-nitro-2,4,6-trimethylbenzene, 4-bromo-2-nitromesitylene, 4-bromo-1,3,5-trimethyl-2-nitrobenzene, 2-bromanyl-1,3,5-trimethyl-4-nitro-benzene
2-bromo-1,3,5-trimethyl-4-nitrobenzene

Specifications

90561-85-6
106°C to 107°C (1.3 mmHg)
MFCD00015924
1958703
3-bromo-2,4,6-trimethylnitrobenzene, 2-bromo-4-nitro-1,3,5-trimethylbenzene, 1-bromo-3-nitro-2,4,6-trimethylbenzene, 4-bromo-2-nitromesitylene, 4-bromo-1,3,5-trimethyl-2-nitrobenzene, 2-bromanyl-1,3,5-trimethyl-4-nitro-benzene
CC1=CC(=C(C(=C1[N+](=O)[O-])C)Br)C
244.088
244.09
2-Bromo-1,3,5-trimethyl-4-nitrobenzene
56°C to 62°C
C9H10BrNO2
1 g
Light sensitive
IUFDRRDWXUSFBG-UHFFFAOYSA-N
2-bromo-1,3,5-trimethyl-4-nitrobenzene
2757200
≥90%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only