missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Bromo-1,3-dichlorobenzene, 97%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals L1106206

 View more versions of this product

Catalog No. AAL1106206


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

2-Bromo-1,3-dichlorobenzene
59°C to 62°C
>100°C (212°F)
MFCD00000574
1681054
UWOIDOQAQPUVOH-UHFFFAOYSA-N
2-bromo-1,3-dichlorobenzene
29568
97%
19393-92-1
241°C to 243°C
C6H3BrCl2
5 g
1-bromo-2,6-dichlorobenzene, 2,6-dichlorobromobenzene, 1,3-dichloro-2-bromobenzene, benzene, 2-bromo-1,3-dichloro, 2,6-dichloro-1-bromobenzene, benzene, 1-bromo-2,6-dichloro, pubchem3610, acmc-209exg, 2,6-dichlorobromo benzene, 2,6-dichlorophenyl bromide
C1=CC(=C(C(=C1)Cl)Br)Cl
225.894
225.91

Chemical Identifiers

19393-92-1
225.894
UWOIDOQAQPUVOH-UHFFFAOYSA-N
29568
C1=CC(=C(C(=C1)Cl)Br)Cl
C6H3BrCl2
MFCD00000574
1-bromo-2,6-dichlorobenzene, 2,6-dichlorobromobenzene, 1,3-dichloro-2-bromobenzene, benzene, 2-bromo-1,3-dichloro, 2,6-dichloro-1-bromobenzene, benzene, 1-bromo-2,6-dichloro, pubchem3610, acmc-209exg, 2,6-dichlorobromo benzene, 2,6-dichlorophenyl bromide
2-bromo-1,3-dichlorobenzene
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

DOTInformation : Hazard Class: 6.1; Packaging Group: III

EINECSNumber : 243-018-6

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only