Learn More
2-Bromo-1,3-diethylbenzene, 94%, Thermo Scientific Chemicals
$114.51 - $319.58
Chemical Identifiers
CAS | 65232-57-7 |
---|---|
Molecular Formula | C10H13Br |
Molecular Weight (g/mol) | 213.12 |
MDL Number | MFCD00017770 |
InChI Key | FPWGIABDOFXETH-UHFFFAOYSA-N |
Synonym | 2,6-diethylbromobenzene, 1-bromo-2,6-diethylbenzene, 2,6-diethyl bromobenzene, benzene, 2-bromo-1,3-diethyl, acmc-20ao3d, 2,6-diethylphenylbromide, 2,6-diethylphenyl bromide, 1,3-diethyl-2-bromobenzene, 2-bromo-1,3-diethyl-benzene, # |
PubChem CID | 522160 |
IUPAC Name | 2-bromo-1,3-diethylbenzene |
SMILES | CCC1=CC=CC(CC)=C1Br |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
AAL0348203
|
Thermo Scientific Chemicals
L0348203 |
1 g |
Each for $114.51
|
|
||||||
AAL0348206
|
Thermo Scientific Chemicals
L0348206 |
5 g |
Each for $319.58
|
|
||||||
Description
2-Bromo-1,3-diethylbenzene is used as raw material in synthetic organic chemistry.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2-Bromo-1,3-diethylbenzene is used as raw material in synthetic organic chemistry.
Solubility
Insoluble in water.
Notes
Incompatible with strong oxidizing agents.
Chemical Identifiers
65232-57-7 | |
213.12 | |
FPWGIABDOFXETH-UHFFFAOYSA-N | |
522160 | |
CCC1=CC=CC(CC)=C1Br |
C10H13Br | |
MFCD00017770 | |
2,6-diethylbromobenzene, 1-bromo-2,6-diethylbenzene, 2,6-diethyl bromobenzene, benzene, 2-bromo-1,3-diethyl, acmc-20ao3d, 2,6-diethylphenylbromide, 2,6-diethylphenyl bromide, 1,3-diethyl-2-bromobenzene, 2-bromo-1,3-diethyl-benzene, # | |
2-bromo-1,3-diethylbenzene |
Specifications
65232-57-7 | |
78°C to 80°C (2 mmHg) | |
1.544 | |
1 g | |
2,6-diethylbromobenzene, 1-bromo-2,6-diethylbenzene, 2,6-diethyl bromobenzene, benzene, 2-bromo-1,3-diethyl, acmc-20ao3d, 2,6-diethylphenylbromide, 2,6-diethylphenyl bromide, 1,3-diethyl-2-bromobenzene, 2-bromo-1,3-diethyl-benzene, # | |
FPWGIABDOFXETH-UHFFFAOYSA-N | |
2-bromo-1,3-diethylbenzene | |
522160 | |
94% |
1.264 | |
C10H13Br | |
MFCD00017770 | |
2501180 | |
Insoluble in water. | |
CCC1=CC=CC(CC)=C1Br | |
213.12 | |
213.12 | |
2-Bromo-1,3-diethylbenzene |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only