missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Bromo-1-[4-(3-bromothien-2-yl)phenyl]ethanone, 97%, Thermo Scientific™

Chemical Identifiers

CAS 944450-89-9
Molecular Formula C12H8Br2OS
Molecular Weight (g/mol) 360.06
MDL Number MFCD09879969
InChI Key YTVAARIMMJZZIE-UHFFFAOYSA-N
Synonym 2-bromo-1-4-3-bromothien-2-yl phenyl ethanone, 2-bromo-1-4-3-bromothiophen-2-yl phenyl ethan-1-one, 2-bromo-1-4-3-bromothiophen-2-yl phenyl ethanone, 4-3-bromothien-2-yl phenacyl bromide, 4-4-bromothien-2-yl phenacyl bromide
PubChem CID 24229752
IUPAC Name 2-bromo-1-[4-(3-bromothiophen-2-yl)phenyl]ethanone
SMILES BrCC(=O)C1=CC=C(C=C1)C1=C(Br)C=CS1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
CC67805CB
SDS View Documents
Thermo Scientific Chemicals
CC67805CB
250mg Amber Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC67805DA
SDS View Documents
Thermo Scientific Chemicals
CC67805DA
1g Amber Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

944450-89-9
360.06
YTVAARIMMJZZIE-UHFFFAOYSA-N
24229752
BrCC(=O)C1=CC=C(C=C1)C1=C(Br)C=CS1
C12H8Br2OS
MFCD09879969
2-bromo-1-4-3-bromothien-2-yl phenyl ethanone, 2-bromo-1-4-3-bromothiophen-2-yl phenyl ethan-1-one, 2-bromo-1-4-3-bromothiophen-2-yl phenyl ethanone, 4-3-bromothien-2-yl phenacyl bromide, 4-4-bromothien-2-yl phenacyl bromide
2-bromo-1-[4-(3-bromothiophen-2-yl)phenyl]ethanone

Specifications

944450-89-9
100.0
Amber Glass Bottle
MFCD09879969
2-bromo-1-4-3-bromothien-2-yl phenyl ethanone, 2-bromo-1-4-3-bromothiophen-2-yl phenyl ethan-1-one, 2-bromo-1-4-3-bromothiophen-2-yl phenyl ethanone, 4-3-bromothien-2-yl phenacyl bromide, 4-4-bromothien-2-yl phenacyl bromide
BrCC(=O)C1=CC=C(C=C1)C1=C(Br)C=CS1
360.06
360.07
2-Bromo-1-[4-(3-bromothien-2-yl)phenyl]ethanone
96.5
97%
C12H8Br2OS
250mg
YTVAARIMMJZZIE-UHFFFAOYSA-N
2-bromo-1-[4-(3-bromothiophen-2-yl)phenyl]ethanone
24229752
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye damage.
Harmful if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes severe skin burns and eye damage.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specifica

GHS Signal Word: Danger

SDS
missing translation for 'documents'

missing translation for 'documents'