missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing
2-Bromo-1-(4-morpholinophenyl)-1-ethanone, 97%, Thermo Scientific™
Chemical Identifiers
CAS |
210832-85-2 |
Molecular Formula |
C12H14BrNO2 |
Molecular Weight (g/mol) |
284.15 |
MDL Number |
MFCD03783555 |
InChI Key |
OUGMZFJPRSTGMJ-UHFFFAOYSA-N |
Synonym |
2-bromo-1-4-morpholinophenyl-1-ethanone, 2-bromo-1-4-morpholinophenyl ethanone, 2-bromo-1-4-morpholin-4-ylphenyl ethanone, 2-bromo-1-4-4-morpholinyl phenyl ethanone, 2-bromo-1-4-morpholin-4-yl-phenyl ethanone, 2-bromo-1-4-morpholin-4-yl phenyl ethanone, 2-bromo-1-4-morpholin-4-ylphenyl ethan-1-one, pubchem23338, acmc-1cqxm, alpha-bromo-4'-morpholinoacetophenone |
PubChem CID |
2795357 |
SMILES |
BrCC(=O)C1=CC=C(C=C1)N1CCOCC1 |
View More Specs
Products
1
Catalog Number |
Mfr. No. |
Quantity |
Price |
Quantity |
|
|
|
|
CC17405CB
SDS
View Documents
|
Thermo Scientific Chemicals
CC17405CB
|
250 mg |
|
|
Add to cart
|
|
|
|
Description
Specifications
Chemical Identifiers
|
210832-85-2 |
|
284.15 |
|
OUGMZFJPRSTGMJ-UHFFFAOYSA-N |
|
2795357 |
|
C12H14BrNO2 |
|
MFCD03783555 |
|
2-bromo-1-4-morpholinophenyl-1-ethanone, 2-bromo-1-4-morpholinophenyl ethanone, 2-bromo-1-4-morpholin-4-ylphenyl ethanone, 2-bromo-1-4-4-morpholinyl phenyl ethanone, 2-bromo-1-4-morpholin-4-yl-phenyl ethanone, 2-bromo-1-4-morpholin-4-yl phenyl ethanone, 2-bromo-1-4-morpholin-4-ylphenyl ethan-1-one, pubchem23338, acmc-1cqxm, alpha-bromo-4'-morpholinoacetophenone |
|
BrCC(=O)C1=CC=C(C=C1)N1CCOCC1 |
Specifications
|
210832-85-2 |
|
C12H14BrNO2 |
|
250 mg |
|
OUGMZFJPRSTGMJ-UHFFFAOYSA-N |
|
2-bromo-1-(4-morpholin-4-ylphenyl)ethanone |
|
2795357 |
|
95% |
|
95% |
|
MFCD03783555 |
|
2-bromo-1-4-morpholinophenyl-1-ethanone, 2-bromo-1-4-morpholinophenyl ethanone, 2-bromo-1-4-morpholin-4-ylphenyl ethanone, 2-bromo-1-4-4-morpholinyl phenyl ethanone, 2-bromo-1-4-morpholin-4-yl-phenyl ethanone, 2-bromo-1-4-morpholin-4-yl phenyl ethanone, 2-bromo-1-4-morpholin-4-ylphenyl ethan-1-one, pubchem23338, acmc-1cqxm, alpha-bromo-4'-morpholinoacetophenone |
|
BrCC(=O)C1=CC=C(C=C1)N1CCOCC1 |
|
284.15 |
|
284.15 |
|
2-Bromo-1-(4-morpholinophenyl)-1-ethanone |
Videos
SDS
missing translation for 'documents'