missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Bromo-1-(4-pentylphenyl)ethan-1-one, 95%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals BTB12841EA

 View more versions of this product

Catalog No. BTB12841EA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

Specifications

Specifications

2-Bromo-1-(4-pentylphenyl)ethan-1-one
94.5
95%
C13H17BrO
MFCD00218837
2-bromo-1-4-pentylphenyl ethan-1-one, 2-bromo-1-4-pentylphenyl ethanone, 2-bromo-4'-pent-1-yl acetophenone, ethanone,2-bromo-1-4-pentylphenyl, 2-bromo-1-4-pent-1-yl phenyl ethan-1-one
CCCCCC1=CC=C(C=C1)C(=O)CBr
269.182
269.18
64328-68-3
100.0
Amber Glass Bottle
CH3(CH2)4C6H4C(O)CH2Br
10g
AVYNDJWQMUOSJZ-UHFFFAOYSA-N
2-bromo-1-(4-pentylphenyl)ethanone
2775670
95%

Chemical Identifiers

64328-68-3
269.182
AVYNDJWQMUOSJZ-UHFFFAOYSA-N
2775670
CCCCCC1=CC=C(C=C1)C(=O)CBr
C13H17BrO
MFCD00218837
2-bromo-1-4-pentylphenyl ethan-1-one, 2-bromo-1-4-pentylphenyl ethanone, 2-bromo-4'-pent-1-yl acetophenone, ethanone,2-bromo-1-4-pentylphenyl, 2-bromo-1-4-pent-1-yl phenyl ethan-1-one
2-bromo-1-(4-pentylphenyl)ethanone
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

SDS Documents

Documents

Promotions

Promotions