missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Bromo-1-(5-methyl-1-phenyl-1H-pyrazol-4-yl)-1-ethanone, ≥95%, Thermo Scientific™

$306.26 - $306.26

Chemical Identifiers

CAS 137577-00-5
Molecular Formula C12H11BrN2O
Molecular Weight (g/mol) 279.137
MDL Number MFCD02681921
InChI Key VYGXRQSIPNGJNK-UHFFFAOYSA-N
Synonym 2-bromo-1-5-methyl-1-phenyl-1h-pyrazol-4-yl-1-ethanone, 2-bromo-1-5-methyl-1-phenylpyrazol-4-yl ethanone, 2-bromo-1-5-methyl-1-phenyl-1h-pyrazol-4-yl ethan-1-one, ethanone, 2-bromo-1-5-methyl-1-phenyl-1h-pyrazol-4-yl, acmc-1c040, 1-phenyl-4-2-bromoacetyl-5-methylpyrazole, 4-bromoacetyl-5-methyl-1-phenyl-1h-pyrazole, 2-bromo-1-5-methyl-1-phenyl-1h-pyrazol-4-yl ethanone, 1-1-phenyl-5-methyl-1h-pyrazole-4-yl-2-bromoethanone, 2-bromo-1-5-methyl-1-phenyl-1h-pyrazol-4-yl-ethanone
PubChem CID 2776439
IUPAC Name 2-bromo-1-(5-methyl-1-phenylpyrazol-4-yl)ethanone
SMILES CC1=C(C=NN1C2=CC=CC=C2)C(=O)CBr
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
CC15505DA
SDS View Documents
Thermo Scientific Chemicals
CC15505DA
1 g
Each for $306.26
 
Description

Description

Specifications

Chemical Identifiers

137577-00-5
279.137
VYGXRQSIPNGJNK-UHFFFAOYSA-N
2776439
CC1=C(C=NN1C2=CC=CC=C2)C(=O)CBr
C12H11BrN2O
MFCD02681921
2-bromo-1-5-methyl-1-phenyl-1h-pyrazol-4-yl-1-ethanone, 2-bromo-1-5-methyl-1-phenylpyrazol-4-yl ethanone, 2-bromo-1-5-methyl-1-phenyl-1h-pyrazol-4-yl ethan-1-one, ethanone, 2-bromo-1-5-methyl-1-phenyl-1h-pyrazol-4-yl, acmc-1c040, 1-phenyl-4-2-bromoacetyl-5-methylpyrazole, 4-bromoacetyl-5-methyl-1-phenyl-1h-pyrazole, 2-bromo-1-5-methyl-1-phenyl-1h-pyrazol-4-yl ethanone, 1-1-phenyl-5-methyl-1h-pyrazole-4-yl-2-bromoethanone, 2-bromo-1-5-methyl-1-phenyl-1h-pyrazol-4-yl-ethanone
2-bromo-1-(5-methyl-1-phenylpyrazol-4-yl)ethanone

Specifications

137577-00-5
100.0
Amber glass bottle
MFCD02681921
2-bromo-1-5-methyl-1-phenyl-1h-pyrazol-4-yl-1-ethanone, 2-bromo-1-5-methyl-1-phenylpyrazol-4-yl ethanone, 2-bromo-1-5-methyl-1-phenyl-1h-pyrazol-4-yl ethan-1-one, ethanone, 2-bromo-1-5-methyl-1-phenyl-1h-pyrazol-4-yl, acmc-1c040, 1-phenyl-4-2-bromoacetyl-5-methylpyrazole, 4-bromoacetyl-5-methyl-1-phenyl-1h-pyrazole, 2-bromo-1-5-methyl-1-phenyl-1h-pyrazol-4-yl ethanone, 1-1-phenyl-5-methyl-1h-pyrazole-4-yl-2-bromoethanone, 2-bromo-1-5-methyl-1-phenyl-1h-pyrazol-4-yl-ethanone
CC1=C(C=NN1C2=CC=CC=C2)C(=O)CBr
279.137
279.14
2-Bromo-1-(5-methyl-1-phenyl-1H-pyrazol-4-yl)-1-ethanone
94.5
95%
C12H11BrN2O
1 g
VYGXRQSIPNGJNK-UHFFFAOYSA-N
2-bromo-1-(5-methyl-1-phenylpyrazol-4-yl)ethanone
2776439
95%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

SDS
missing translation for 'documents'

missing translation for 'documents'