missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Bromo-1-indanol 98.0+%, TCI America™
$46.60 - $184.54
Chemical Identifiers
| CAS | 5400-80-6 |
|---|---|
| Molecular Formula | C9H9BrO |
| Molecular Weight (g/mol) | 213.07 |
| MDL Number | MFCD00003798 |
| InChI Key | RTESDSDXFLYAKZ-UHFFFAOYNA-N |
| Synonym | 2-bromo-1-indanol, 2-bromoindan-1-ol, 1-hydroxy-2-bromoindan, 2-bromo-1-hydroxyindane, 2-bromo-1-hydroxyhydrindene, 1h-inden-1-ol, 2-bromo-2,3-dihydro, trans-2-bromo-1-indanol, bromoindanol, acmc-20ajgx, pubchem3783 |
| PubChem CID | 95444 |
| IUPAC Name | 2-bromo-2,3-dihydro-1H-inden-1-ol |
| SMILES | OC1C(Br)CC2=CC=CC=C12 |
Chemical Identifiers
| 5400-80-6 | |
| 213.07 | |
| RTESDSDXFLYAKZ-UHFFFAOYNA-N | |
| 95444 | |
| OC1C(Br)CC2=CC=CC=C12 |
| C9H9BrO | |
| MFCD00003798 | |
| 2-bromo-1-indanol, 2-bromoindan-1-ol, 1-hydroxy-2-bromoindan, 2-bromo-1-hydroxyindane, 2-bromo-1-hydroxyhydrindene, 1h-inden-1-ol, 2-bromo-2,3-dihydro, trans-2-bromo-1-indanol, bromoindanol, acmc-20ajgx, pubchem3783 | |
| 2-bromo-2,3-dihydro-1H-inden-1-ol |
Specifications
| 5400-80-6 | |
| C9H9BrO | |
| 5 g | |
| RTESDSDXFLYAKZ-UHFFFAOYNA-N | |
| 2-bromo-2,3-dihydro-1H-inden-1-ol | |
| 95444 | |
| ≥98.0% (GC) | |
| 2-Bromo-1-indanol |
| 131°C | |
| MFCD00003798 | |
| 2-bromo-1-indanol, 2-bromoindan-1-ol, 1-hydroxy-2-bromoindan, 2-bromo-1-hydroxyindane, 2-bromo-1-hydroxyhydrindene, 1h-inden-1-ol, 2-bromo-2,3-dihydro, trans-2-bromo-1-indanol, bromoindanol, acmc-20ajgx, pubchem3783 | |
| OC1C(Br)CC2=CC=CC=C12 | |
| 213.07 | |
| 213.07 | |
| Crystalline Powder |
Safety and Handling
EINECSNumber : (4)-1771
TSCA : No