missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2-Bromo-1-propene 97.0+%, TCI America™
Supplier: TCI America B126910G
Specifications
2-Bromo-1-propene | |
Yellow | |
C3H5Br | |
10 g | |
2-bromopropene, isopropenyl bromide, 1-propene, 2-bromo, 2-bromo-1-propene, 2-bromopropylene, propene, 2-bromo, isopropylene bromide, alpha-methylvinyl bromide, .alpha.-methylvinyl bromide, 2-bromo-propene | |
CC(=C)Br | |
120.977 | |
120.98 | |
Liquid |
557-93-7 | |
48°C | |
MFCD00000140 | |
1993 | |
PHMRPWPDDRGGGF-UHFFFAOYSA-N | |
2-bromoprop-1-ene | |
11202 | |
≥97.0% (GC) |
Chemical Identifiers
557-93-7 | |
120.977 | |
PHMRPWPDDRGGGF-UHFFFAOYSA-N | |
11202 | |
CC(=C)Br |
C3H5Br | |
MFCD00000140 | |
2-bromopropene, isopropenyl bromide, 1-propene, 2-bromo, 2-bromo-1-propene, 2-bromopropylene, propene, 2-bromo, isopropylene bromide, alpha-methylvinyl bromide, .alpha.-methylvinyl bromide, 2-bromo-propene | |
2-bromoprop-1-ene |
Safety and Handling
RTECSNumber : UC7085000
TSCA : Yes
Recommended Storage : Refrigerator