Learn More
2-Bromo-2-butene, 98%, mixture of cis and trans, Thermo Scientific Chemicals
$150.23 - $150.23
Chemical Identifiers
CAS | 13294-71-8 |
---|---|
Molecular Formula | C4H7Br |
Molecular Weight (g/mol) | 135.00 |
MDL Number | MFCD00000141 |
InChI Key | UILZQFGKPHAAOU-ONEGZZNKSA-N |
Synonym | 2-bromo-2-butene, trans-2-bromo-2-butene, 2-butene, 2-bromo, e-2-bromobut-2-ene, 2-bromo-2-butene cis, e-2-bromo-2-butene, 1-methyl-1-propenyl bromide, cis-2-bromo-2-butene, e-2-bromo-but-2-ene, 2e-2-bromo-2-butene # |
PubChem CID | 5364387 |
IUPAC Name | (E)-2-bromobut-2-ene |
SMILES | C\C=C(/C)Br |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | |||||
AC201510250
|
Thermo Scientific Chemicals
201510250 |
25 mL | Glass bottle |
Each for $150.23
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
13294-71-8 | |
135.00 | |
UILZQFGKPHAAOU-ONEGZZNKSA-N | |
5364387 | |
C\C=C(/C)Br |
C4H7Br | |
MFCD00000141 | |
2-bromo-2-butene, trans-2-bromo-2-butene, 2-butene, 2-bromo, e-2-bromobut-2-ene, 2-bromo-2-butene cis, e-2-bromo-2-butene, 1-methyl-1-propenyl bromide, cis-2-bromo-2-butene, e-2-bromo-but-2-ene, 2e-2-bromo-2-butene # | |
(E)-2-bromobut-2-ene |
Specifications
13294-71-8 | |
100.0 | |
1.3200g/mL | |
<1°C | |
97.5% min. (GC) | |
C4H7Br | |
CH3CH=C(Br)CH3 | |
25 mL | |
1.32 | |
Solubility in water: immiscible | |
C\C=C(/C)Br | |
135.00 | |
135 | |
Liquid |
97.5 | |
Brown to Yellow | |
82°C to 90°C (740.0 mmHg) | |
Authentic | |
Glass bottle | |
1.4590 to 1.462 | |
MFCD00000141 | |
01, 205 | |
2-bromo-2-butene, trans-2-bromo-2-butene, 2-butene, 2-bromo, e-2-bromobut-2-ene, 2-bromo-2-butene cis, e-2-bromo-2-butene, 1-methyl-1-propenyl bromide, cis-2-bromo-2-butene, e-2-bromo-but-2-ene, 2e-2-bromo-2-butene # | |
UILZQFGKPHAAOU-ONEGZZNKSA-N | |
(E)-2-bromobut-2-ene | |
5364387 | |
98% | |
2-Bromo-2-butene, 98% |
Safety and Handling
GHS H Statement:
Highly flammable liquid and vapor.
GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
EINECSNumber : 236-316-2
TSCA : TSCA
Recommended Storage : Product may darken during storage
RUO – Research Use Only