Learn More
2-Bromo-2'-hydroxyacetophenone, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals B2572206
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
2-Bromo-2'-hydroxyacetophenone | |
45°C to 47°C | |
MFCD01727570 | |
UN3263 | |
SGPKEYSZPHMVNI-UHFFFAOYSA-N | |
2-bromo-1-(2-hydroxyphenyl)ethanone | |
200671 | |
97% |
2491-36-3 | |
C8H7BrO2 | |
5 g | |
2-bromo-2'-hydroxyacetophenone, 2-bromo-1-2-hydroxyphenyl ethanone, 2-hydroxyphenacyl bromide, ethanone, 2-bromo-1-2-hydroxyphenyl, 2-bromo-1-2-hydroxyphenyl ethan-1-one, acetophenone, 2-bromo-2'-hydroxy-6ci,7ci,8ci, ethanone,2-bromo-1-2-hydroxyphenyl, 2-bromoacetyl phenol, acmc-1cg90, 4-08-00-00329 beilstein handbook reference | |
C1=CC=C(C(=C1)C(=O)CBr)O | |
215.046 | |
215.05 |
Chemical Identifiers
2491-36-3 | |
215.046 | |
SGPKEYSZPHMVNI-UHFFFAOYSA-N | |
2-bromo-1-(2-hydroxyphenyl)ethanone |
C8H7BrO2 | |
MFCD01727570 | |
200671 | |
C1=CC=C(C(=C1)C(=O)CBr)O |
Safety and Handling
GHS H Statement
H314-H318-H302
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
H302-H314-H335
DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, BASIC, ORGANIC, N.O.S.
RTECSNumber : KM5556400
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only