missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Bromo-3,3,3-trifluoro-1-propene, 97%, Thermo Scientific Chemicals

$106.88 - $106.88

Chemical Identifiers

CAS 1514-82-5
Molecular Formula C3H2BrF3
Molecular Weight (g/mol) 174.95
MDL Number MFCD00077469
InChI Key QKBKGNDTLQFSEU-UHFFFAOYSA-N
Synonym 2-bromo-3,3,3-trifluoropropene, 2-bromo-3,3,3-trifluoro-1-propene, 1-propene, 2-bromo-3,3,3-trifluoro, 2-bromo-3,3,3-trifluoro-propene, 2-bromo-3,3,3-trifluoro-prop-1-ene, 3,3,3-trifluoro-2-bromo-1-propene, 2-bromotrifluoropropene, acmc-1bt5r, ksc495o1d, 3,3,3-trifluoro-2-bromopropene
PubChem CID 272696
IUPAC Name 2-bromo-3,3,3-trifluoroprop-1-ene
SMILES FC(F)(F)C(Br)=C
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC434460010
SDS View Documents
Thermo Scientific Chemicals
434460010
1 g Glass bottle N/A
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

1514-82-5
174.95
QKBKGNDTLQFSEU-UHFFFAOYSA-N
272696
FC(F)(F)C(Br)=C
C3H2BrF3
MFCD00077469
2-bromo-3,3,3-trifluoropropene, 2-bromo-3,3,3-trifluoro-1-propene, 1-propene, 2-bromo-3,3,3-trifluoro, 2-bromo-3,3,3-trifluoro-propene, 2-bromo-3,3,3-trifluoro-prop-1-ene, 3,3,3-trifluoro-2-bromo-1-propene, 2-bromotrifluoropropene, acmc-1bt5r, ksc495o1d, 3,3,3-trifluoro-2-bromopropene
2-bromo-3,3,3-trifluoroprop-1-ene

Specifications

1514-82-5
100.0
1.6500g/mL
Authentic
Glass bottle
1.3520 to 1.3550 (20°C, 589nm)
1 g
2-bromo-3,3,3-trifluoropropene, 2-bromo-3,3,3-trifluoro-1-propene, 1-propene, 2-bromo-3,3,3-trifluoro, 2-bromo-3,3,3-trifluoro-propene, 2-bromo-3,3,3-trifluoro-prop-1-ene, 3,3,3-trifluoro-2-bromo-1-propene, 2-bromotrifluoropropene, acmc-1bt5r, ksc495o1d, 3,3,3-trifluoro-2-bromopropene
FC(F)(F)C(Br)=C
174.95
174.95
Liquid
96.0
Colorless
34°C
96% min. (GC)
C3H2BrF3
MFCD00077469
1.65
QKBKGNDTLQFSEU-UHFFFAOYSA-N
2-bromo-3,3,3-trifluoroprop-1-ene
272696
97%
2-Bromo-3, 3, 3-trifluoro-1-propene
Videos
Safety and Handling

Safety and Handling

GHS H Statement
May cause respiratory irritation.
May cause drowsiness or dizziness.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER or doctor/physician if you feel unwell.

GHS Signal Word: Warning

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only