missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2-Bromo-3,3,3-trifluoro-1-propene 97.0+%, TCI America™
Supplier: TCI America B18015G
Specifications
2-Bromo-3,3,3-trifluoro-1-propene | |
Yellow | |
C3H2BrF3 | |
5 g | |
QKBKGNDTLQFSEU-UHFFFAOYSA-N | |
2-bromo-3,3,3-trifluoroprop-1-ene | |
272696 | |
≥97.0% (GC) |
1514-82-5 | |
34°C | |
MFCD00077469 | |
2-bromo-3,3,3-trifluoropropene, 2-bromo-3,3,3-trifluoro-1-propene, 1-propene, 2-bromo-3,3,3-trifluoro, 2-bromo-3,3,3-trifluoro-propene, 2-bromo-3,3,3-trifluoro-prop-1-ene, 3,3,3-trifluoro-2-bromo-1-propene, 2-bromotrifluoropropene, acmc-1bt5r, ksc495o1d, 3,3,3-trifluoro-2-bromopropene | |
FC(F)(F)C(Br)=C | |
174.95 | |
174.95 | |
Liquid |
Chemical Identifiers
1514-82-5 | |
174.95 | |
QKBKGNDTLQFSEU-UHFFFAOYSA-N | |
272696 | |
FC(F)(F)C(Br)=C |
C3H2BrF3 | |
MFCD00077469 | |
2-bromo-3,3,3-trifluoropropene, 2-bromo-3,3,3-trifluoro-1-propene, 1-propene, 2-bromo-3,3,3-trifluoro, 2-bromo-3,3,3-trifluoro-propene, 2-bromo-3,3,3-trifluoro-prop-1-ene, 3,3,3-trifluoro-2-bromo-1-propene, 2-bromotrifluoropropene, acmc-1bt5r, ksc495o1d, 3,3,3-trifluoro-2-bromopropene | |
2-bromo-3,3,3-trifluoroprop-1-ene |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator