missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Bromo-3-chloro-5-(trifluoromethyl)pyridine, 97%, Thermo Scientific™

Chemical Identifiers

CAS 75806-84-7
Molecular Formula C6H2BrClF3N
Molecular Weight (g/mol) 260.438
MDL Number MFCD00153072
InChI Key SMTKGMYGLYWNDL-UHFFFAOYSA-N
Synonym 2-bromo-3-chloro-5-trifluoromethyl pyridine, librarion l938, abbypharma ap-13-5059, pyridine, 2-bromo-3-chloro-5-trifluoromethyl, pubchem3000, acmc-1bicf, ksc498c5d, buttpark 153\33-66, 2-bromo-3-chloro-5 trifluoromethyl pyridine
PubChem CID 2736237
IUPAC Name 2-bromo-3-chloro-5-(trifluoromethyl)pyridine
SMILES C1=C(C=NC(=C1Cl)Br)C(F)(F)F
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
BTB09453DA
SDS View Documents
Thermo Scientific Chemicals
BTB09453DA
1g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
BTB09453DE
SDS View Documents
Thermo Scientific Chemicals
BTB09453DE
5g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
BTB09453EA
SDS View Documents
Thermo Scientific Chemicals
BTB09453EA
10g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

75806-84-7
260.438
SMTKGMYGLYWNDL-UHFFFAOYSA-N
2736237
C1=C(C=NC(=C1Cl)Br)C(F)(F)F
C6H2BrClF3N
MFCD00153072
2-bromo-3-chloro-5-trifluoromethyl pyridine, librarion l938, abbypharma ap-13-5059, pyridine, 2-bromo-3-chloro-5-trifluoromethyl, pubchem3000, acmc-1bicf, ksc498c5d, buttpark 153\33-66, 2-bromo-3-chloro-5 trifluoromethyl pyridine
2-bromo-3-chloro-5-(trifluoromethyl)pyridine

Specifications

75806-84-7
100.0
Amber glass bottle
MFCD00153072
2-bromo-3-chloro-5-trifluoromethyl pyridine, librarion l938, abbypharma ap-13-5059, pyridine, 2-bromo-3-chloro-5-trifluoromethyl, pubchem3000, acmc-1bicf, ksc498c5d, buttpark 153\33-66, 2-bromo-3-chloro-5 trifluoromethyl pyridine
C1=C(C=NC(=C1Cl)Br)C(F)(F)F
260.438
260.44
2-Bromo-3-chloro-5-(trifluoromethyl)pyridine
96.5
97%
C6H2BrClF3N
1g
SMTKGMYGLYWNDL-UHFFFAOYSA-N
2-bromo-3-chloro-5-(trifluoromethyl)pyridine
2736237
97%
Videos
Safety and Handling

Safety and Handling

GHS P Statement Causes skin irritation. Causes serious eye irritation.

GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF ON SKIN: Wash with plenty of soap and water. Take off contaminated clothing and wash before reuse. IF IN EYES: Rinse cautio

Warning

SDS
missing translation for 'documents'

missing translation for 'documents'