missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Bromo-3-methylaniline, ≥97%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals H6401403

 View more versions of this product

Catalog No. AAH6401403


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

2-Bromo-3-methylaniline
Brown
48°C (0.03 mmHg)
C7H8BrN
MFCD06411367
UN2810
benzenamine, 2-bromo-3-methyl, 3-amino-2-bromotoluene, 2-bromo-3-methylbenzenamine, 2-bromo-3-methyl-benzenamine, 2-bromo-3-methylphenylamine, 2-bromo-3-methyl-phenylamine, acmc-20af7i, 2-bromo-3-methyl aniline, ksc602o1n, benzenamine,2-bromo-3-methyl
CC1=C(C(=CC=C1)N)Br
186.052
186.05
Liquid
54879-20-8
1.489 g/mL
>110°C (230°F)
1.605
1 g
2079474
VJNUZLYTGSGDHR-UHFFFAOYSA-N
2-bromo-3-methylaniline
603781
≥97%

Chemical Identifiers

54879-20-8
186.052
VJNUZLYTGSGDHR-UHFFFAOYSA-N
603781
CC1=C(C(=CC=C1)N)Br
C7H8BrN
MFCD06411367
benzenamine, 2-bromo-3-methyl, 3-amino-2-bromotoluene, 2-bromo-3-methylbenzenamine, 2-bromo-3-methyl-benzenamine, 2-bromo-3-methylphenylamine, 2-bromo-3-methyl-phenylamine, acmc-20af7i, 2-bromo-3-methyl aniline, ksc602o1n, benzenamine,2-bromo-3-methyl
2-bromo-3-methylaniline
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302+H312+H332-H315-H319-H335

DOTInformation : Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.

TSCA : No

storageNote1 : 2° to 8°C

Recommended Storage : Keep cold

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only